Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
The new Cole-Parmer catalog has over 20 pages of dataloggers and data recorders to monitor temperture, humidity, and other variables. Request yours today.
Featured Products
Compact USB Data Loggers
compact USB datalogger
  • Small compact design
  • Includes software and battery
  • Monitor Temperature, Humidity, Voltage, and Current
from $63.00 More details
Temperature Data Logger
Temperature datalogger
  • Large backlit LCD
  • Front panel keypad
  • On-screen statistics
from $499.00 More details
High Temperature Data Logger
high temperature data recorder
  • Miniature Size
  • Fully Submersible
  • 316L Stainless Steel Enclosure
from $499.00 More details
Temperature/Humidity Paperless Recorder
temperature and humidity paperless data recorder
  • Paperless recorder measures temperature, humidity plus calculates dew point
  • Internal memory records up to 49,000 data points
  • Unit features audible and visual alarms with hi/low setpoints
from $229.99 More details
Cole-Parmer Catalog > Data Loggers
Dataloggers Technology
A data logger is an electronic instrument that records information over a period of time for later use. The type of information recorded is determined by the user, for example, such physical parameters as temperature, relative humidity, pressure, voltage, water level, etc. Cole-Parmer stocks all of these data recording devices and there are various types, depending on your application, to choose from.
With the recent introduction of new technology, it has become possible to build miniature electronic devices that are battery operated and can automatically record information for later retrieval by a computer. To minimize cost and size, full control and operation of the data logger is achieved through the use of a personal computer with data logging software. The data logger will collect measurements, such as temperature, relative humidity, pressure, voltage, water level, etc. at periodic intervals and record them in chronological order within the data logger's memory for later retrieval. Data Loggers vary in accuracy and functionality. Cole-Parmer offers numerous types of data recorder devices used in today's Industrial, Chemical Process, Food & Beverage, Laboratory, and Healthcare/Pharaceutical fields.
Temperature
Cole-Parmer offers a wide variety of Temperature Data Recorders for monitoring and recording temperature in a myriad of applications. The manufacturer's software easily downloads data to your computer to display and analyze data in degrees of Celsius, Fahrenheit, Kelvin or Rankine.
Temperature & Humidity
Cole-Parmer offers a full line of Temperature & Humidity Data Loggers with a wide range and high accuracy, these humidity/temperature recorders are ideal for most humidity applications. The manufacturer's software will display data in units of temperature, relative humidity, dew point, and water vapor concentration in standard or metric units.
Need help in selecting the best data logger for your specific application? Let our highly trained Application Specialists provide you with additional product specifications, guide you through data logger selection and maintenance, and offer support throughout the operational life of the data logger you purchase. They are thoroughly schooled on the products we sell and can answer the most challenging questions.
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams