Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Choose from over 250 pages of products for life science, inlcuding 14 pages of electrophoresis products. Choose from a number of well known brands including Corning, Thermo Scientific, UVP, EMD Chemicals, Nalge NUNC, and Eppendorf. Request yours today.
Electrophoresis
Cole-Parmer offers a complete line of products for electrophoresis including horizontal and vertical gel electrophoresis systems, reagents, imaging systems, and power supplies. Along with our wide range of products, we also offer expert assistance in finding the right product. For more information or help choosing the right product for your need, contact our Application Specialists.
Gel electrophoresis is a method by which DNA, RNA or proteins are separated by electric current typically in an agarose gel. Agarose gels are cast and placed in a gel box with buffer. Sample is pipetted into wells cast into the gels and the electric current is applied to the system. The electric current causes the molecules to move through the gel at different rates depending on their mass and mass to charge ratio. Once complete, staining of the molecules within the gel allow for visibility by the researcher. If the molecules are able to fluoresce, an image of the gel and molecular movement can be taken.
Applications
Electrophoresis Products
Browse the NEW Life Science Products & Solutions Catalog (pdf)
Life Science solutions
  • Electrophoresis & Western Blotting
  • Lab Equipment & Supplies
  • Cole-Parmer
    625 East Bunker Court
    Vernon Hills, Illinois 60061
    Phone: 1-800-323-4340
    Fax: 1-847-247-2929
    E-Mail: info@Coleparmer.com
    Copyright © 2010
    Cole-Parmer Sites  Feedback
    Cole-Parmer:
    United States
    Canada
    China
    United Kingdom
    India
    McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams