Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
88 pages of flow meters and hundreds of pages of flow accessories including pumps, fittings, tubing and process control. Request yours today.
Featured Products
Handheld Doppler Flowmeter
handheld doppler flowmeter
  • Uses advanced signal processing to measure flow in pipes sized 1
  • Range 0.30 to 30 ft/sec (0.1 to 9m/sec)
  • For use with fluids with particulates or bubbles; 100 ppm, 100 microns or larger
from $1635.00 More details
Portable Doppler Flowmeter
portable doppler flowmeter
  • Ideal for measuring flow rates of dirty or particulate ridden fluids
  • Programmable models display velcoity in ft/sec, m/sec, flow rate in GPM, cu ft.sec, GPD and L/sec
  • Available with a 4 to 20mA output signal
from $3438.00 More details
Cole-Parmer Ultrasonic Hybrid Doppler/Transit Time Flowmeters
hybrid doppler/transit-time ultrasonic flowmeter
  • Noninvasive technology works with both clean or dirty fluids
  • RS-232, RS-485, Ethernet, and USB connections to real-time user PC software
  • NEMA 4X (IP66) enclosure is weather-resistant for outdoor use, and includes carrying case
from $3500.00 More details
Cole-Parmer Doppler Ultrasonic Flowmeter
doppler ultrasonic flowmeter
  • In-field linearization allows adaptation to a wide variety of applications
  • Accuracy is ±2% full-scale over calibrated span
  • Nonintrusive, clamp-on transducers fit pipes from 1/4
from $1630.00 More details
Cole-Parmer Catalog > Flow Meters > Ultrasonic Flow Meters
Ultrasonic Flowmeters
Common Applications for Ultrasonic Flow Meters
  • Gas-stack flow measurement in power plant scrubbers
  • Cryogenic liquids (time transit ultrasonic flow meters only)
  • Influent and effluent water flow (doppler flow meters only)
  • Waste water (doppler flow meters only)
  • Light to medium crude oils in the petroleum refining industry (time transit meters only)
  • Makeup water
  • Pure and ultra-pure fluids in semiconductor, pharmaceutical, and the food & beverage industries (time transit ultrasonic flow meters only)
  • Digester feed control (doppler flow meters only)
  • Acids and liquefied gases in the chemical industry
  • Cooling water
  • Water distribution systems used in agriculture and irrigation
  • Clarifier monitoring
Cole-Parmer's range of ultrasonic flow meters and doppler flowmeters offers process contamination free measurement of liquids and gas flow. A typical ultrasonic flow meter will consist of a transducer that is mounted on to the exterior of the pipe and an indicator or totalizer to display the output. This can easily be installed without disturbing existing pipe work. Most ultrasonic flow meters will work with pipe sizes between 1/2" and 200" diameter. There are two main types of ultrasonic flow meters; doppler flow meters and time transit ultrasonic flow meters.
A doppler flow meter requires particulates or bubbles in the media. The minimum diameter size of the particulate is typically 30 microns and requires a minimum concentration levels of 25 ppm. Since some meters may require slightly larger concentrations of particulate, it is a good idea to check the specifications table. Doppler flow meters are mainly used for liquid applications (roughly 88%) while the rest are used for gas (11%) and steam (1%) applications.
A time transit ultrasonic flow meter requires a clean liquid without particulates or bubbles and can be used for both liquid and gas applications. It has better accuracy than that of a doppler flow meter and will typically offer accuracies of ±2% full scale.
Why Use an Ultrasonic Flow Meter
  • Doppler flow meters are suitable for slurries
  • Suitable for aggressive chemicals as no contact with the media
  • Allows for easy installation without disturbing existing pipe work
  • Process contamination free measurement of flow
  • No leak potential
Limitations of Ultrasonic Flow Meters
  • Higher initial setup costs than other flow technologies
  • Pipe material must be compatible with ultrasonic sensor
  • Requires sufficient knowledge of the flow and media to give some confidence in readings
To find the right flow meter for your application use Cole-Parmer's on-line flow meter quick search. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs.
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams