Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 25 pages of fume hoods, extractors, enclosures, and accessories. Request your copy today. Request yours today.
Featured Products
Cole-Parmer® Compact Fume Hoods
Cole-Parmer<reg> Compact Fume Hoods
  • NEW LOWER PRICE!!
  • Economical and efficient system for your basic fume hood needs
  • Units come Complete with dished composite resin work surface, vaporproof light fixture, and built-in exhaust blower
from $4541.00 More details
Labconco®47- and 70-Inch Basic Fume Hoods
47-inch fiberglass fume hood with standard blower
  • Perfect when you need a full-sized ventilated hood to handle small quantities of chemicals
  • These fume hoods are an ideal alternative to traditional fume hoods
from $3630.00 More details
Labconco®Protector® XStreamTM Laboratory Hoods
4-Foot XStream Hood, 115V, without fixtures
  • Lab safety that pays for itself!
  • Combines the finest elements of design and construction
  • XStream's are low face-velocity hoods that are safe, efficient, and extremely durable
from $7280.00 More details
Cole-Parmer Catalog > Fume Hoods
Fume Hoods
Cole-Parmer offers a wide variety of fume hoods for every laboratory application including general purpose hoods, vented enclosures, ducted fume hoods, biosafety and PCR enclosures, and extractor systems. Choose from many trusted names including Labconco, Thermo Scientific Hamilton, and Misonix..
Need assistance in choosing the right fume hood for your needs? Contact our Application Specialists for further assistance.
Types of Fume Hoods
Balance Enclosures provide user protection by keeping powders, particulates and fumes contained during weighing procedures. Balance Enclosures may be ducted to the outside or connected to a HEPA and/or carbon filtered exhaust back into the laboratory.
Ductless Fume Hoods are enclosures that pass contaminated air through a filter(s) before returning air to the room. Use for locations where outside ventilation can not be achieved. Can eliminate very high percentages of particulates and biological hazards. Select airborne chemical levels can be reduced to safe levels as verified with monitoring protocols.
Ducted Fume Hoods have ventilation to outside provides the most rigorous user protection, particularly for chemically dangerous fumes. Many employ "by-pass" designs, which enable consistent airflows in the enclosure for various sash positions. Low airflow designs provide user protection while minimizing energy costs.
Biological Safety Cabinets provide effective containment for working with human pathogens. BSC cabinets have three classes as sedignated by the National Sanitation Foundation—Class I, II, and III, with some sub class levels.
BSC Class I have an un-recirculated airflow away from the lab worker that, after going through a HEPA filter, is vented into the room. They provide the lab worker medium protection but do not protect the samples in the bacinet from contamination.
BCS Class II should be used if a lab worker is using microorganisms in containment levels 2, 3, and 4 labs. These cabinets are divided into two types—A and B, determined by construction, airflow velocities, patterns, and exhaust systems. They protect the lab worker, samples, and the external environment.
BCS Class III have both HEPA filtered supply and exhaust air and are totally gas tight and enclosed. Tests are performed while using long sleeved gloves. The cabinet is under negative pressure of at least 120 Pa while the exterior exhaust system maintains airflow. This protects the lab worker, samples, and the external environment.
PCR Enclosures are designed specifically to privide a Class 5 work space for PCR experiments by moving HEPA filtered air downward. PCR enclosures are unique in that they have a UV light that eliminates DNA and RNA contaminates
Extractor Systems provide localized ventilation at the contaminant source. Extractor arms can be easily positioned to remove fumes or air-borne particulates. Often used when the fume source can not be readily moved to a conventional hood or when fumes are innocuous, e.g. bad-smelling without being chemically dangerous. May vent to outside or recirculate into the room through a filter.
Don't forget all our fume hood Accessories, including monitors and alarms, blowers, base stands and cabinets, prefilters, sash stops, and vinyl curtains.
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams