Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 20 pages of infrared thermometers. Choose from our wide selection to meet the demands of any application. Request yours today.
Featured Products
Oakton InfraPro Infrared Thermometers
Oakton IR Thermometers
  • Measure hazardous targets from a distance
  • Rugged enough for industrial use yet compact
  • Single-point laser targets the measurement area
from $199.00 More details
Fluke® HVACPro 561® Infrared Thermometer
Fluke  ®  HVACPro 561
  • Designed for the HVAC market, with features that can be used in any IR application.
  • Single-point laser targets the measurement area
  • Unique emissivity selector makes it easy for anyone to use
from $165.00 More details
Oakton Mini-InfraPro Infrared Thermometers
Oakton Mini-InfraPro
  • Anywhere that temperature is a factor, a Oakton IR thermometer should be close at hand
  • Pistol-grip handle is comfortable and easy to use
  • Measure surface temperatures quickly and accurately
from $75.00 More details
Fluke 63, 66, 68 566, 568 Infrared Thermometers
Fluke  568
  • Ideal for maintenance diagnostics in demanding environments
  • Laser targets your measurement area
  • For special surfaces such as concrete all Fluke models have adjustable emissivity (except Fluke 63)
from $379.00 More details
Oakton Mini TempTestr Infrared Thermometer
Oakton Mini TempTestr
  • Economical infrared thermometer fits right in your pocket
  • Features minimum/maximum memory readings and Hold function
  • Measure surface temperatures quickly and accurately
from $32.00 More details
Oakton Food Safety Infrared Thermometer
Oakton Food Safety Infrared Thermometer
  • A combination contact/noncontact thermometer for all food applications
  • Go/no-go LEDs provide quick check of HACCP food zones
  • Sealed IP54 housing make for easy cleaning
from $169.00 More details
Cole-Parmer Catalog > Temperature > Infrared Thermometers
Infrared Thermometers
Just Point and Shoot
Oakton Infrared Thermometer
When Noncontact Readings are Critical,
Cole-Parmer has the Infrared Thermometer for the Job!
Measuring surface temperature can provide clues to such problems as leaky ductwork, over-stressed motors, faulty equipment, or food that is in danger of spoiling. There is no easier way to take such readings than with a Cole-Parmer handheld infrared thermometer! Ideal for use when objects are extremely hot, moving quickly, difficult to reach, or vulnerable to contamination. Cole-Parmer offers handheld infrared thermometers to meet the demands of any application. The Cole-Parmer®, Oakton®, Fluke® and other brands are industry leaders in accuracy, durability, and high optical resolution. Our products are backed by expert applications assistance and customer service.
Lightweight. Compact.
Quick and Easy to Use.
Noncontact infrared (IR) thermometers use infrared technology to quickly and conveniently measure an objects surface temperature. They provide fast, accurate, repeatable readings without ever touching the target. Just aim, pull the trigger, and read the temperature on the LCD.
  • Safely measure hot, hazardous, or hard-to-reach surfaces
  • Measure food temperature without contaminating or damaging product
  • Log several readings per second instead of waiting several minutes for a single reading from a contact thermometer
Quick and Handy
Take Repeatable Readings
with Mini Infrared Thermometers
Designed to Fit in the Palm of Your Hand
  • Ideal for fast inspections in food preparation, life sciences, and field use
  • Quick temperature measurement from less than 12 inches—take readings of any liquid, solid, or semisolid in less than one second
  • Our most economical units
  • Models small enough to fit in a shirt pocket
  • Adjustable emissivity available
Food Safety
Use Food Safety IR Thermometers
to Meet GMPs and HACCP
Requirements as set by the FDA
  • Measure and log food temperatures during cooking, serving, cooling, storage,transport, and reheating to increase product quality and consistency
  • Quickly and accurately monitor for HACCP danger zones
  • Measure temperatures up to 626°F (330°C) for pathogen reduction
  • Reduce liability and waste by preventing cross-contamination
  • Available with IP65- or IP54-rated housings for easy cleaning
  • Ideal for food preparation areas, labs, and production plants
General-Purpose
Make Quick, Basic Temperature Checks
with Compact IR Thermometers
  • Economically priced for distance readings up to 20 feet
  • Large, easy-to-read LCD allows rapid, multiple readings
  • Ideal for automotive repair, plant maintenance, and HVAC equipment monitoring
  • Some models are available with thermocouple input for the ability to gather internal or direct contact measurements
  • Rugged enough for industrial environments
Industrial Professional
Get the Job Done Right the First Time with
InfraPro® Professional-Grade IR Guns
  • Designed for the professional technician
  • Broader temperature range and improved optics allow accurate measuring from greater distances
  • Ideal for use around moving parts, and for dangerous or difficult-to-reach objects
  • Monitor air stratification, furnace performance, electrical panels and connectors, and more
  • Available with an RTD input for direct contact measurements with a temperature range up to 1400°F (760°C)
Precision Measurement
Take High-Accuracy Readings at Distances up to 60 Feet
and Greater with These Precise Infrared Thermometers
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams