Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 30 pages on various laboratory tubing formulations along with hundreds of pages on fittings, flowmeters and other fluid handling products Request yours today.
Featured Products
Cole-Parmer PVC Lab Tubing
Flexible PVC lab tubing
  • A high-quality tubing for general purpose and laboratory use
  • Resistant to oxidation, most chemicals and bacteria
  • Available in multiple sizes to meet your specific needs
from $13.00 More details
Pure-Fit® Tru-Valve for Flexible Tubing
Tru-Valve
  • Unique design may be used on existing lines and systems
  • Sturdy glass-reinforced nylon construction
  • Easy to custom control flow through simple threaded handle
from $20.50 More details
Lab Tubing Racks
Laboratory Tubing racks
  • Help organize up to 10 tubing lines!
  • Racks available with adhesive backing or screw mounts for secure mounting
  • Select sizes may separate to custom-sized retainers
from $16.00 More details
Cole-Parmer Catalog > Lab Tubing
Laboratory Tubing
Cole-Parmer offers one of the most extensive ranges of laboratory tubing anywhere! We offer so many alternatives in flexible and rigid tubing that we can supply the best tubing for virtually any fluid handling need. Whether for basic or harsh chemicals, sensitive biological reactions, higher pressures or temperatures, or simple fluid transfers, we carry a lab tubing to meet your needs. Consider the range of parameters we can meet with our tubing products:
  • Tubing Size with inside diameters as small as 0.002" (0.05 mm) to 2"(50.8mm)
  • Pressure ratings up to 1000 psi (69 bar)
  • Tubing to withstand Vacuum level of 29.9" of mercury (Hg) at 70°F (23° C)
  • Temperature ratings up to 550° F (288° C)
  • Chemical compatible laboratory tubing for research, testing, food/beverage, pharmaceutical, and chemical needs, with many formulations suitable for aggressive chemicals and solvents. Excellent in applications requiring transfer tubing, dispensing tubing, flexible tubing, rigid tubing, or any other type of plastic tubing or rubber tubing.
  • Tubing suitable for sterilization techniques such as gamma irradiation, autoclaving and ethylene oxide
  • Tubing Material Certification are available free of charge at time of order for a wide selection of our tubing. The certification states which regulatory standards the tubing material meets.
  • Quantities for general use as well as bulk spools of up to 500 ft (150 m) for some popular formulations.
Choose from several different types of laboratory tubing:
Flexible tubing
Peroxide-Cured Silicone Tubing
  • Flexible, soft tubing available in more than 50 formulations, including Tygon®, silicones, PVC and vinyls, C-FLEX®, PharMed®, rubbers (including Norprene®), polyurethanes, Flexelene, and Hubtron
  • Able to withstand temperatures up to 450° F (232° C)
  • Sizes as small as 0.008 inch (0.19 mm) ID
  • Pressures from vacuum to 450 psi (31 bar)
Microbore laboratory tubing
Microbore EVA tubing
  • 8 formulations of specialty microbore tubing, including Tygon® formulations, Viton®, silicone, PTFE, and ethyl vinyl acetate (EVA), among others
  • Sizes as fine as human hair
  • Perfect for low-flow applications or precision pumping and dispensing
Rigid laboratory tubing
PTFE Tubing
  • Flexible, soft tubing available in more than 20 formulations, including fluoropolymers (PTFE, PFA, FEP, PVDF, and ETFE)®, nylons, polyethylenes, polycarbonate, polypropylene, polyimide, Bev-A-Line® formulations, PEEK, and metals (stainless steels, copper, and nickel)
  • Able to withstand temperatures up to 550° F (288° C)
  • Sizing as small as 0.010 inch (0.25 mm) ID
  • Pressures from vacuum to over 11,000 psi (760 bar)
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams