Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Meet your most stringent quality control needs...
Cole-Parmer® features these all new Atago® refractometers and polarimeter.
From determining concentrations of soluble solids to measuring refractive index and percent Brix of foods or beverages- Cole-Parmer carries the Atago product for your specific application. Measuring the physical properties of pharmaceuticals, cosmetics, industrial coolants and chemicals is simple with these new Atago products, providing precision accuracy you need, with the name you can trust - Atago.
Digital "Hand-Held" Pocket Brix Refractometer
MM-02941-51 PAL-1
  • Portable and Economical
  • Measurement range of 0.0% to 53.0% Brix.
  • Rugged IP65 design is ideal for field use and easy cleanup.
Automatic Refractometer
MM-81100-40 SMART-1
  • Choose from four scales - inverted sugar, Brix, HFCS-55 or HFCS-42.
  • 0.05% accuracy with a measurement range of 0.00% to 95.00% Brix!
Programmable Digital Refractometer
MM-81100-30 RX-7000alpha
  • Measure refractive index samples from 1.3250 to 1.7000.
  • Built in thermo module ensures precision accuracy.
  • No water bath circulator required!
Automatic Polarimeter
MM-81200-15 AP-100
  • Fully automatic, precision accuracy with the touch of a button!
  • Measure the specific optical rotation or the concentration of optically active substances.
Other Atago Refractometers and Polarimeters
Products and News for...