Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Purchase any of these Digi-Sense thermometers, and receive a
FREE Tech Clip Stopwatch
Digi-Sense Type J, K, or T Thermocouple Thermometers and RTD Thermometer
Make sure you use the "UZ" code in front of the item number in your shopping cart to get the FREE TechClip. (Example: UZ-91100-00).
  • Used with Cole-Parmer Thermocouple or RTD probes
  • Used in food processing plants, science labs, agriculture, building maintenance and more!
  • IP-54 rated housing; splash-and dust-resistant
  • Ergonomically designed
  • Easy-to-use automatic field calibration ensures accurate readings
  • Intrinsically safe, UL-listed
  • Hold function
  • View minimum/maximum temperature
  • Can be factory calibrated to NIST standard (optional; order with purchase)
UZ-91100-00 Digi-Sense Type J thermocouple thermometer
UZ-91100-10 Digi-Sense Type K thermocouple thermometer
UZ-91100-20 Digi-Sense Type T thermocouple thermometer
UZ-93410-00 Digi-Sense RTD thermocouple thermometer with three-pin connector
Order probes separately
Thermocouple probes
RTD probes
Tech Clip Stopwatch/Keychain
Shock-resistant stopwatch features:
  • Built-in dial compass
  • °F thermometer
  • Selectable 12/24 hour clock with date, day of week, and month
  • 24-hour stopwatch
  • Selectable audible alarm
  • Sturdy keychain clip
Yours FREE with the purchase of a Digi-Sense Thermometer!