Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
       
       
Home Products & Services Technical Library myCP Account Contact/About Us
Product Categories
Virtual Print Catalog
Special Offers
What's New
Free Catalogs/Literature
Free eNews
Bioconnect Custom Bioprocess Assemblies
InnoCal Calibration Services
Custom Ordering Solutions
Technical Library Index
Chemical Compatibility Database
Parametric Searches
Product Selection Guides
Product Maintenance
FAQs
Application Assistance
MSDS Search
Instruction Manual Search
Online Unit Converters
Articles and White Papers
myCP Home
my Orders
myCP Profile
myCP Addresses
my Shopping Lists
my Quick Lists
my Projects
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help




Quick Order!
Item # Qty






Feedback



Electrochemistry featured brands- WTW and Corning    

pH Meters
General Accessories


pH Meters  [back to top]
 Corning M500-Series Benchtop pH Meters
EW-58907-20 - CORNING ADVANCED MODEL BENCHTOP PH METER, 115 VAC
  Corning Waterproof pH Meters and Kits
EW-58904-00 - Corning waterproof pH/mV/temperature meter, ATC
 WTW Benchtop pH Meter Kits
EW-58901-12 - WTW pH meter, 110 VAC
  WTW VARIO Waterproof pH Meter
EW-58908-05 - WTW Vario pH meter w/ SenTix 21 Electrode
 WTW Waterproof pH Meter Kits
EW-58889-00 - WTW Waterproof pH Meter Kit
General Accessories  [back to top]
 General Accessories and Replacement Items
EW-58900-50 - Replacement pH electrode with ATC, waterproof, for meter kits 58901-00 and -05
EW-58905-60 - Three-in-one pH/reference/temperature electrode, epoxy body, BNC for meters 58910-00 to -15