| Evaporators [back to top] |
|   |  | IKA Rotary Evaporators |
| EW-28710-40 - IKA CONTROL ROTARY EVAPORATOR WITH VERTICAL UNCOATED GLASSWARE 115VAC |
| EW-28710-45 - IKA CONTROL ROTARY EVAPORATOR WITH VERTICAL SAFETY COATED GLASSWARE 115VAC |
| EW-28710-50 - IKA CONTROL ROTARY EVAPORATOR WITH DIAGONAL UNCOATED GLASSWARE 115VAC |
| EW-28710-55 - IKA CONTROL ROTARY EVAPORATOR WITH DIAGONAL SAFETY COATED GLASSWARE 115VAC |
| Homogenizers [back to top] |
|   |  | IKA T 10 Basic Ultra Turrax Homogenizer |
| EW-04720-70 - IKA T-10 BASIC HOMOGENIZER 115 VAC |
| EW-04720-71 - IKA T-10 BASIC HOMOGENIZER 230 VAC |
|   |  | IKA T 10 basic ULTRA-TURRAX Homogenizer Accessories |
| EW-04720-72 - DISPERSING ELEMENT 5MM X 75 MM FOR IKA T-10 BASIC HOMOGENIZER |
| EW-04720-73 - DISPERSING ELEMENT 8MM X 95MM FOR IKA T-10 BASIC HOMOGENIZER |
| EW-04720-74 - DISPERSING ELEMENT 10MM X 100MM FOR IKA T-10 BASIC HOMOGENIZER |
| EW-04720-75 - PLASTIC DISPERSING ELEMENT 7MM X 65MM (12/PK) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-76 - PLASTIC DISPERSING ELEMENT 7MM X 65MM (25/PK) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-77 - PLASTIC DISPERSING ELEMENT 7MM X 110 MM (12/PK) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-78 - PLASTIC DISPERSING ELEMENT 7MM X 110MM (25/PK) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-79 - STAND (R 104) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-80 - SUPPORT CLAMP (R 200) FOR T-10 BASIC HOMOGENIZER |
| EW-04720-81 - BOSS HEAD CLAMP (H 44) FOR T-10 BASIC HOMOGENIZER |
|   |  | IKA T-25 Homogenizer Rotor-Stator Generators |
| EW-04719-52 - 50 to 2500 mL sample volume range, 200 mm L x 25 mm OD, coarse dispersion type, Aqueous and Organic Solution |
| EW-04719-53 - 100 to 2000 mL sample volume range, 200 mm L x 25 mm OD, fine dispersion type, Aqueous and Organic solution |
| EW-04720-10 - 1 to 50 mL sample volume range, 200 mm L x 10 mm OD, coarse dispersion type, Aqueous and Organic solution |
| EW-04720-11 - 0.3 to 5 mL sample volume range, 100 mm L x 8 mm OD, coarse dispersion type, Aqueous and Organic solution |
| EW-04720-12 - 10 to 500 mL sample volume range, 200 mm L x 18 mm OD, coarse dispersion type, Aqueous and Organic solution |
| EW-04720-90 - PLASTIC DISPERSING ELEMENT 10MM X 150MM (5/PK) FOR IKA T-25 DIGITAL HOMOGENIZER |
| EW-04720-91 - PLASTIC DISPERSING ELEMENT 10MM X 150MM (10/PK) FOR IKA T-25 DIGITAL HOMOGENIZER |
| EW-04720-92 - PLASTIC DISPERSING ELEMENT 14MM X 150MM (5/PK) FOR IKA T-25 DIGITAL HOMOGENIZER |
| EW-04720-93 - PLASTIC DISPERSING ELEMENT 14MM X 150MM (10/PK) FOR IKA T-25 DIGITAL HOMOGENIZER |
| EW-04720-99 - CLOSED TUBE FOR IKA T-18 AND T-25 HOMOGENIZERS, 50 ML (DISPOSABLE) |
|   |  | IKA ULTRA TURRAX Tube Disperser Sample Tubes |
| EW-04720-95 - STIRRING TUBE 50 ML, (10/PK) |
| EW-04720-96 - DISPERSION TUBE 50 ML, (10/PK) |
| EW-04720-97 - BALL MILL TUBE 50 ML, GLASS (10/PK) |
| EW-04720-98 - BALL MILL TUBE 50 ML, CERAMIC (10/PK) |
| EW-04738-01 - STIRRING TUBE 15 ML, (10/PK) |
| EW-04738-02 - DISPERSION TUBE 15 ML, (10/PK) |
| EW-04738-03 - BALL MILL TUBE 15 ML, GLASS (10/PK) |
| EW-04738-04 - BALL MILL TUBE 15 ML, CERAMIC (10/PK) |
|   |  | IKA T-25 Digital High-Speed Homogenizer Systems |
| EW-04739-01 - IKA T-25 ULTRA-TURRAX Digital Homogenizer |
| EW-04739-03 - IKA T-25 ULTRA-TURRAX Digital Homogenizer Kit, 115VAC |
|   |  | IKA ULTRA TURRAX Tube Dispersers |
| EW-04738-89 - IKA ULTRA TURRAX Tube Drive, Blue Line 115 VAC |
| Hot Plates [back to top] |
|   |  | IKA Ceramic-Top Hot Plates |
| EW-04671-55 - ETS-D5 Temperature Probe |
| EW-04671-60 - IKA Ceramic Hot Plate, 4" x 4", 115v |
| EW-04671-62 - IKA Ceramic Hot Plate, 4" x 4", 230V |
| EW-04671-64 - IKA Ceramic Hot Plate, 7" x 7", 115v |
| EW-04671-66 - IKA Ceramic Hot Plate, 7" x 7", 230V |
| EW-04671-68 - IKA Ceramic Hot Plate, 10" x 10", 115v |
| EW-04671-70 - IKA Ceramic Hot Plate, 10" x 10", 230v |
| Mills [back to top] |
|   |  | IKA Continuous Feed Grinding Drive |
| EW-04301-30 - IKA Continuous feed grinding drive, 115 VAC/60 Hz |
| EW-04301-35 - IKA Continuous feed grinding drive, 230 VAC/50 Hz |
|   |  | IKA Continuous Feed Grinding Heads |
| EW-04300-30 - IKA Cutting-grinding head |
| EW-04300-32 - IKA Impact grinding head |
|   |  | IKA Economical Analytical Mill |
| EW-04300-10 - IKA Economical Analytical Mill, 115 VAC, 50/60 Hz |
|   |  | IKA Heavy-Duty Analytical Mill |
| EW-04301-10 - IKA Heavy-Duty Analytical Mill, 115 VAC/60 Hz |
| EW-04301-15 - IKA Heavy-Duty Analytical Mill, 230 VAC/50 Hz |
| Mixers and Overhead Stirrers [back to top] |
|   |  | IKA EUROSTAR Electronic Overhead Stirrers |
| EW-50704-50 - IKA EUROSTAR Power Control-Visc P4, 115 VAC |
| EW-50704-55 - IKA EUROSTAR Power Control-Visc P4, 230 VAC |
| EW-50704-60 - IKA EUROSTAR Power Control-Visc P7, 115 VAC |
| EW-50704-65 - IKA EUROSTAR Power Control-Visc P7, 230 VAC |
|   |  | IKA High-Viscosity Mixers |
| EW-50703-00 - IKA High-Viscosity Mixer, 115 VAC |
| EW-50703-05 - IKA High-Viscosity Mixer, 230 VAC |
| EW-50703-10 - IKA High-Viscosity Mixer with Digital Display, 115 VAC |
| EW-50703-15 - IKA High-Viscosity Mixer with Digital Display, 230 VAC |
|   |  | IKA Lab Egg Compact Mixer |
| EW-50702-04 - IKA Lab Egg Compact Mixer, Blue |
|   |  | IKA Mixer Accessories |
| EW-50702-55 - IKA Compact Mixer, Stirring Element |
| EW-50703-25 - Propeller stirrer, 4-bladed |
| EW-50703-26 - Propeller stirrer, 4-bladed |
| EW-50703-30 - Propeller stirrer, 3-bladed |
| EW-50703-31 - Propeller stirrer, 3-bladed |
| EW-50703-32 - Propeller stirrer, 3-bladed |
| EW-50703-33 - Propeller stirrer, 3-bladed |
| EW-50703-34 - Propeller stirrer, 3-bladed, PTFE-Coated |
| EW-50703-35 - Turbine Stirrer |
| EW-50703-36 - Turbine Stirrer |
| EW-50703-37 - Turbine Stirrer |
| EW-50703-40 - Dissolver stirrer, 3 1/8" |
| EW-50703-41 - Dissolver stirrer, 4" |
| EW-50703-42 - Dissolver stirrer, 1 5/8" |
| EW-50703-45 - Centrifugal stirrer |
| EW-50703-46 - Centrifugal stirrer |
| EW-50703-50 - Paddle stirrer, 2 3/4" |
| EW-50703-51 - Paddle stirrer, 2 3/4" |
| EW-50703-52 - Paddle stirrer 5 7/8" |
| EW-50703-55 - Anchor stirrer, 1 3/4" |
| EW-50703-56 - Anchor stirrer, 3 1/2" |
| EW-50703-57 - Anchor stirrer, PTFE-coated, 2 3/8" |
| EW-50703-58 - Anchor stirrer, 5 7/8" |
| EW-50703-60 - Kneading stirrer |
| EW-50703-65 - Paddle stirrer, vane |
| EW-50703-70 - Strap clamp |
| EW-50703-71 - Strap clamp |
| EW-50703-75 - Flexible coupling |
| EW-50703-85 - IKA Remote control |
| EW-50703-90 - Mixer plate stand, 22" |
| EW-50703-91 - Mixer plate stand, 31 1/2" |
| EW-50703-92 - Mixer plate stand, 39 1/3" |
| EW-50703-93 - Mixer H-stand, 39 3/4" |
| EW-50703-94 - Mixer H-stand, adjsutable to 39 3/4" |
| EW-50703-96 - Boss Head Clamp |
| EW-50703-97 - Boss Head Clamp |
| EW-50703-98 - Boss Head Clamp |
| Shakers [back to top] |
|   |  | Fixing Clips |
| EW-04303-88 - 1000-mL fixing clip for shaker platform |
| EW-51505-92 - Fixing Clips for IKA Orbital Shakers, 50 mL |
| EW-51505-95 - Fixing Clips for IKA Orbital Shakers, 500 mL |
|   |  | IKA Lab Dancer Compact Shaker |
| EW-84001-80 - IKA Lab Dancer Compact Shaker, 115V |
| EW-84001-81 - IKA Lab Dancer Compact Shaker, 230V |
|   |  | IKA Variable-Speed Digital Orbital Shakers |
| EW-04303-00 - IKA Variable-Speed Digital Orbital Shaker, Basic model, 230V |
| EW-04303-02 - IKA Variable-Speed Digital Orbital Shaker, Basic model, 115V |
| EW-04303-04 - IKA Variable-Speed Digital Orbital Shaker, Control model, 230V |
| EW-04303-06 - IKA Variable-Speed Digital Orbital Shaker, Control model, 115V |
|   |  | IKA Incubating Shakers |
| EW-51715-50 - Universal Platform with Clamping Rolls for IKA KS 4000 Incubated Shakers |
| EW-51715-52 - Flask Clamp Platform for IKA KS 4000 Incubated Shakers |
| EW-51715-54 - Platform with Rubber Mat Base for IKA KS 4000i Incubated Shakers |
| EW-51715-72 - STICKMAX Self-Adhesive Shaker Mat |
|   |  | IKA Orbital Shakers |
| EW-04304-00 - IKA Basic Orbital Shaker, 115v |
| EW-04304-02 - IKA Basic Orbital Shaker, 230v |
| EW-04304-04 - IKA Digital Orbital Shaker, 115v |
| EW-04304-06 - IKA Digital Orbital Shaker, 230v |
| Stirrers [back to top] |
|   |  | IKA Big Squid Stirrer |
| EW-84001-90 - Ika Big Squid Stirrer 115V |
| EW-84001-92 - Ika Big Squid Stirrer 230V |
|   |  | IKA Digital Ceramic-Top Stirrers and Stirring Hot Plates |
| EW-04671-26 - IKA Ceramic Stirring Hot Plate, 4" x 4", 115v |
| EW-04671-28 - IKA Ceramic Stirring Hot Plate, 4" x 4", 230V |
| EW-04671-30 - IKA Digital Stirring Hot Plate, 115v |
| EW-04671-32 - IKA Ceramic Stirring Hot Plate, 230v |
| EW-04671-34 - IKA Ceramic Stirring Hot Plate, 10" x 10", 115v |
| EW-04671-36 - IKA Ceramic Stirring Hot Plate, 10" x 10", 230v |
| EW-04671-38 - IKA Ceramic Stirrer, 4" x 4", 115v |
| EW-04671-40 - IKA Ceramic Stirrer, 4" x 4", 230V |
| EW-04671-42 - IKA Ceramic Stirrer, 7" x 7", 115v |
| EW-04671-44 - IKA Ceramic Stirrer, 7" x 7", 230V |
| EW-04671-46 - IKA Ceramic Stirrer, 10" x 10", 115v |
| EW-04671-48 - IKA Ceramic Stirrer, 10" x 10", 230V |
|   |  | Ika Mini Color Squid Stirrer |
| EW-84001-00 - IKA Mini Color Squid Stirrer 115V |
| EW-84001-02 - Ika Mini Color Squid Stirrer 230V |
|   |  | IKA Ultra-Flat Lab Disc Stirrer |
| EW-84001-70 - IKA Ultra-Flat Lab Disc, 120VAC |
|   |  | Ika Battery-Powered Stirrer |
| EW-84001-08 - Ika Battery-Operated Stirrer |
|   |  | IKA Large Capacity Digital Stirrers |
| EW-04671-82 - IKA Large Capacity Digital Stirrer, 19" x 19", 115V |
| EW-04671-83 - IKA Large Capacity Digital Stirrer, 19" x 19", 230V |
|   |  | IKA RCT and RET basic IKAMAG Round Top Digital Stirring Hot Plates |
| EW-04671-21 - IKA RCT Basic IKAMAG Digital Round-Top Stirring Hot Plate, 115V |
| EW-04671-23 - IKA RCT Basic IKAMAG Digital Round-Top Stirring Hot Plate, 230v |
| EW-04671-25 - RET basic IKAMAG Stirring Hot Plate 115V |
| EW-04671-27 - RET basic IKAMAG stirring hot plate 230V |
|   |  | Large-Volume Stir Bars |
| EW-04627-00 - Large-Volume Stir Bar, 4-1/2"L x 1-1/16"dia |
| General Accessories [back to top] |
|   |  | General Accessories and Replacement Items |
| EW-04300-24 - Tungsten-carbide blade, |
| EW-04300-26 - Star-shaped, 301 SS blade |
| EW-04300-28 - Additional grinding chamber, 304 SS |
| EW-04300-33 - Sieve, 0.25 mm |
| EW-04300-34 - Sieve, 0.5 mm |
| EW-04300-35 - Sieve, 1.0 mm |
| EW-04300-37 - Sieve, 2.0 mm |
| EW-04300-38 - Sieve, 3.0 mm |
| EW-04300-52 - Additional blade, 304 SS |
| EW-04300-54 - Cutting blade, 440 SS for pulverizing soft, fibrous materials |
| EW-04300-56 - Beater, carbide coated 420 SS for pulverizing materials with a hardness of up to 9 Mohs |
| EW-04300-58 - Grinding chamber, 250 mL, 316 SS |
| EW-04303-50 - Universal attachment |
| EW-04303-52 - Fixing clip attachment |
| EW-04303-54 - Dish attachment |
| EW-04303-56 - Test tube support |
| EW-04303-58 - Fixing clip; 20mL |
| EW-04303-60 - Fixing clip; 100mL |
| EW-04303-62 - Fixing clip; 200 mL |
| EW-50704-70 - labworldsoft software |