Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.

  Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register

Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help

Cole-Parmer® Chemicals    


Looking for more? See our complete line of 30,000 Chemicals!


Chemicals  [back to top]
 1,2-Dichloroethane
EW-89000-54 - 1 2-DICHLOROETHANE ACS 4L
 1-Butanol
EW-89000-34 - N-BUTYL ALCOHOL ACS 500mL
EW-89000-35 - N-BUTYL ALCOHOL ACS 4L
 2-Propanol
EW-89000-96 - ISOPROPYL ALCOHOL HPLC 4L
 Acetic Acid
EW-89000-02 - ACETIC ACID GLACIAL ACS 2.5L
 Acetic Acid, Glacial
EW-89000-01 - ACETIC ACID GLACIAL ACS 500mL
EW-89000-03 - ACETIC ACID GLACIAL ACS 2.5L
 Acetone
EW-89000-04 - ACETONE ACS 500mL
EW-89000-06 - ACETONE ACS 500mL
EW-89000-07 - ACETONE ACS 1L
EW-89000-08 - ACETONE ACS 4L
EW-89000-09 - ACETONE ACS 20L
EW-89000-11 - ACETONE HPLC 4L
EW-89000-13 - ACETONE CP 4L
 Acetonitrile
EW-89000-14 - ACETONITRILE ACS 500mL
EW-89000-16 - ACETONITRILE ACS 1L
EW-89000-17 - ACETONITRILE ACS 4L
EW-89000-18 - ACETONITRILE ACS 20L
EW-89000-19 - ACETONITRILE HPLC 4L
  Agar
EW-89000-20 - AGAR 500G
 Alcohol
EW-89100-59 - REAGENT ALCOHOL ABSOLUTE 4L
 Alizarin Red S
EW-89200-12 - ALIZARIN RED S 25G
 Aluminum Nitrate Nonahydrate
EW-89000-21 - ALUMINUM NITRATE 9-HYDRATE ACS 500G
 Aluminum Sulfate Octadecahydrate
EW-89000-22 - ALUMINUM SULFATE HYDRATED ACS 500G
 Ammonium Acetate
EW-89000-23 - AMMONIUM ACETATE CRYSTAL ACS 500G
 Ammonium Bifluoride
EW-89000-24 - AMMONIUM BIFLUORIDE Technical 500G
 Ammonium Chloride
EW-89000-25 - AMMONIUM CHLORIDE ACS 500G
 Ammonium Hydroxide
EW-89000-26 - AMMONIUM HYDROXIDE ACS 500mL
EW-89000-27 - AMMONIUM HYDROXIDE ACS 2.5L
EW-89000-28 - AMMONIUM HYDROX 27% Technical 2.5L
  Ammonium Nitrate
EW-89000-29 - AMMONIUM NITRATE CRYSTAL ACS 500G
 Ascorbic Acid
EW-89000-30 - ASCORBIC ACID CRYSTAL USP 500G
 Barium Chloride Dihydrate
EW-89000-31 - BARIUM CHLORIDE ACS 500G
 Boric Acid
EW-89000-32 - BORIC ACID ACS 500G
EW-89000-33 - BORIC ACID ACS 2.5KG
 Bromothymol Blue Solution
EW-89200-13 - BROMOTHYMOL BLUE 0.04% AQUEOUS SOLUTION 500ML
 Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue)
EW-89200-14 - BUFFER SOLUTION PH 10.00 BLUE 500ML
EW-89200-16 - BUFFER SOLUTION PH 10.00 BLUE 1L
 Buffer, Reference Standard, pH 2.00 ± 0.01 at 25°C
EW-89200-17 - BUFFER SOLUTION PH 2.00 500ML
 Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C (Color Coded Red)
EW-89200-18 - BUFFER SOLUTION PH 4.01 RED 500ML
EW-89200-19 - BUFFER SOLUTION PH 4.01 RED 1L
 Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow)
EW-89200-21 - BUFFER SOLUTION PH 7.00 YELLOW 500ML
EW-89200-22 - BUFFER SOLUTION PH 7.00 YELLOW 1L
 Calcium Carbonate
EW-89000-37 - CALCIUM CARBONATE POWDER ACS 2.5KG
 Calcium Chloride
EW-89000-38 - CALCIUM CHLORIDE ANHYDROUS 500G
 Calcium Chloride Dihydrate
EW-89000-39 - CALCIUM CHLORIDE DIHYDRATE POWDER ACS 500G
EW-89000-40 - CALCIUM CHLORIDE DIHYDRATE POWDER ACS 2.5KG
 Calcium Hydroxide
EW-89000-41 - CALCIUM HYDROXIDE POWDER ACS 500G
 Calcium Nitrate Tetrahydrate
EW-89000-42 - CALCIUM NITRATE CRYSTAL ACS 500G
 Chloroform
EW-89000-44 - CHLOROFORM ACS 500mL
EW-89000-45 - CHLOROFORM ACS 1L
EW-89000-46 - CHLOROFORM ACS 4L
 Chromium (VI) Trioxide
EW-89000-47 - CHROMIUM TRIOXIDE FLAKE ACS 2.5KG
 Citric acid
EW-89000-48 - CITRIC ACID ANHYDROUS ACS 500G
  Cobalt(II) chloride, hexahydrate
EW-89000-49 - COBALT CHLORIDE CRYSTAL Reagent 500G
 Copper (II) Chloride Dihydrate
EW-89000-51 - COPPER CHLORIDE DIHYDRATE PURIFIED Reagent 500G
EW-89000-52 - COPPER CHLORIDE DIHYDRATE PURIFIED Reagent 500G
 Cyclohexane
EW-89000-53 - CYCLOHEXANE ACS 500mL
 EDTA Titrant, 0.100 Molar (M/10)
EW-89200-23 - EDTA TITRANT 0.100 Molar 3.8L
 Ethanol, Anhydrous
EW-89000-58 - ETHANOL, ANHYDROUS DENATURED 200 Proof 1 Gallon
EW-89000-60 - ETHANOL, ANHYDROUS DENATURED 200 Proof 4L
  Ether anhydrous
EW-89000-56 - ETHER ACS 950mL
 Ethyl Alcohol Denatured
EW-89000-57 - ETHANOL, ANHYDROUS DENATURED 200 Proof 500mL
EW-89000-59 - ETHANOL, ANHYDROUS DENATURED 200 Proof 4L
 Ethylene Glycol
EW-89000-61 - ETHYLENE GLYCOL PURIFIED 99% 20L
EW-89000-62 - ETHYLENE GLYCOL Reagent 500mL
EW-89000-63 - ETHYLENE GLYCOL Reagent 4L
 Formaldehyde
EW-89000-65 - FORMALDEHYDE 37% SOLUTION ACS 500mL
EW-89000-66 - FORMALDEHYDE 37% SOLUTION ACS 4L
EW-89000-67 - FORMALDEHYDE 37% SOLUTION ACS 19L
 Formalin, Neutral Buffered
EW-89200-24 - 10% BUFFERED FORMALIN 4L
EW-89200-26 - 10% BUFFERED FORMALIN 20L
 Glycerin
EW-89000-72 - GLYCERIN USP 500mL
 Glycerol
EW-89000-68 - GLYCEROL ACS 500mL
EW-89000-69 - GLYCEROL ACS 4L
EW-89000-71 - GLYCEROL ACS 19L
 Heptane
EW-89000-73 - N-HEPTANE Reagent 1L
EW-89000-74 - N-HEPTANE Reagent 4L
 Hexanes
EW-89000-75 - HEXANES ACS 500mL
EW-89000-76 - HEXANES ACS 4L
EW-89000-77 - HEXANES HPLC 4L
 Hydrochloric Acid
EW-89000-79 - HYDROCHLORIC ACID ACS 500mL
EW-89000-81 - HYDROCHLORIC ACID ACS 500mL
EW-89000-82 - HYDROCHLORIC ACID ACS 2.5L
EW-89000-84 - HYDROCHLORIC ACID Electronic 2.5L
 Hydrochloric Acid Solution
EW-89200-28 - Hydrochlor Acid 0.010N 1L
EW-89200-32 - HYDROCHLORIC ACID 0.100N 1L
EW-89200-33 - HYDROCHLOR ACID 0.50N 1L
EW-89200-37 - HYDROCHLOR ACID 1.0N 3.8L
 Hydrochloric Acid, 0.0200 Normal (N/50)
EW-89200-29 - Hydrochlor Acid 0.020N 1L
EW-89200-30 - Hydrochlor Acid 0.020N 3.8L
EW-89200-31 - Hydrochlor Acid 0.020N 20L
 Hydrochloric Acid, 0.500 Normal (N/2)
EW-89200-34 - HYDROCHLOR ACID 0.50N 3.8L
 Hydrochloric Acid, 1.00 Normal
EW-89200-35 - HYDROCHLOR ACID 1.00N 1L
EW-89200-36 - HYDROCHLOR ACID 1.00N 3.8L
EW-89200-38 - HYDROCHLOR ACID 1.0N 20L
 Hydrochloric Acid, 10% (v/v) Aqueous Solution (1 + 9)
EW-89200-27 - HYDROCHLORIC ACID 10%(v/v) 19L
 Hydrochloric Acid, 6.00 Normal
EW-89200-39 - HYDROCHLORIC ACID 6.0N 1L
EW-89200-40 - HYDROCHLOR ACID 6.0N 3.8L
EW-89200-41 - HYDROCHLOR ACID 6.0N 20L
 Hydrofluoric acid
EW-89000-85 - HYDROFLUORIC ACID 48% ACS 473mL
 Hydrogen Peroxide
EW-89000-86 - HYDROGEN PEROX 3% Reagent 473mL
EW-89000-87 - HYDROGEN PEROXIDE 30% ACS 473mL
EW-89000-88 - HYDROGEN PEROXIDE 30% ACS 4L
 Iodine
EW-89000-89 - IODINE RESUBLIMED CR ACS 500G ACS 500G
 Iron (III) Chloride Anhydrous
EW-89000-64 - FERRIC CHLORIDE ACS 500G
 Isopropanol
EW-89000-90 - ISOPROPYL ALCOHOL ACS 500mL
EW-89000-91 - ISOPROPYL ALCOHOL ACS 1L
EW-89000-92 - ISOPROPYL ALCOHOL ACS 4L
EW-89000-93 - ISOPROPYL ALCOHOL ACS 20L
 Isopropanol, 70% (v/v) Aqueous Solution
EW-89000-99 - ISOPROPYL ALCOHOL 70% USP 3.8L
EW-89100-01 - ISOPROPYL ALCOHOL 70% USP 20L
 Magnesium Carbonate
EW-89100-02 - MAGNESIUM CARBONATE POWDER USP 500G
 Magnesium Chloride Hexahydrate
EW-89100-03 - MAGNESIUM CHLORIDE CRYSTAL ACS 500G
EW-89100-04 - MAGNESIUM CHLORIDE CRYSTAL ACS 2.5KG
  Magnesium Oxide
EW-89100-05 - MAGNESIUM OXIDE LIGHT POWDER USP 500G
 Magnesium Sulfate Anhydrous
EW-89100-06 - MAGNESIUM SULFATE ANHYDROUS POWDER Reagent 50kg
 Methanol
EW-89100-08 - METHANOL, ANHYDROUS ACS 4L
EW-89100-09 - METHANOL, ANHYDROUS ACS 4L
EW-89100-11 - METHANOL USP 4L
EW-89100-12 - METHANOL HPLC 4L
 Methyl alcohol
EW-89100-07 - METHANOL, ANHYDROUS ACS 1L
 Methylene Blue
EW-89200-42 - METHYLENE BLUE CHLORIDE Certifiable Stain 100g
 Methylene Chloride
EW-89100-13 - METHYLENE CHLORIDE ACS 500ML
EW-89100-14 - METHYLENE CHLORIDE ACS 4L
EW-89100-15 - METHYLENE CHLORIDE HPLC 4L
 N,N-Dimethylformamide
EW-89000-55 - N N-DIMETHYLFORMAMIDEACS ACS 500mL
 n-Pentane
EW-89100-24 - N-PENTANE TECHNICAL 4 L CAN Technical 4L
  Nickel (II) Chloride Hexahydrate
EW-89100-16 - NICKEL CHLORIDE Reagent 500G
 Nitric Acid
EW-89100-17 - NITRIC ACID 70% ACS 500mL
EW-89100-18 - NITRIC ACID 70% ACS 2.2L
 o-Phosphoric Acid
EW-89100-35 - PHOSPHORIC ACID 85% ACS 500mL
EW-89100-36 - PHOSPHORIC ACID 85% ACS 2.2L
 Potassium Sulfate
EW-89100-55 - POTASSIUM SULFATE CRYSTAL ACS 2.5KG
 Potassium Bromide
EW-89100-38 - POTASSIUM BROMIDE CRYSTAL ACS 2.5KG
 Oleic acid, sodium salt
EW-89100-22 - OLEIC ACID SODIUM SALT 1G
 Pentane
EW-89100-23 - N-PENTANE Reagent 4L
 Pepsin
EW-89100-25 - PEPSIN POWDER 100g
 Petroleum Ether
EW-89100-26 - PETROLEUM ETHER 35-60C ACS 500mL
EW-89100-28 - PETROLEUM ETHER 35-60C ACS 4L
EW-89100-29 - PETROLEUM ETHER 35-60C ACS 20L
EW-89100-27 - PETROLEUM ETHER 35-60C ACS 950mL
 Phenol
EW-89100-31 - PHENOL 90% LIQUIFIED USP 500ML
 Phenolphthalein
EW-89100-32 - PHENOLPHTHALEIN POWDER USP 500G
EW-89100-33 - PHENOLPHTHALEIN POWDER USP 125G
 Phenolphthalein Indicator, 1% (w/v) in 95% (v/v) Alcohol, Neutralized
EW-89200-43 - PHENOLPHTHALEIN 1% in 95% ALCOHOL 500ML
EW-89200-44 - PHENOLPHTHALEIN 1% in 95% ALCOHOL 1L
EW-89200-45 - PHENOLPHTHALEIN 1% in 95% ALCOHOL 3.8L
EW-89200-46 - PHENOLPHTHALEIN 1% in 95% ALCOHOL 100mL
 Phosphate Buffer, pH 7.2, for Biochemical Oxygen Demand (BOD) Testing
EW-89200-47 - PHOSPHATE BUFFER PH 7.2 1L
 Phosphoric acid
EW-89100-34 - PHOSPHORIC ACID 85% ACS 500mL
 Potassium Bromide
EW-89100-37 - POTASSIUM BROMIDE CRYSTAL ACS 500G
 Potassium Chloride
EW-89100-39 - POTASSIUM CHLORIDE CRYSTAL ACS 500G
EW-89100-40 - POTASSIUM CHLORIDE CRYSTAL ACS 2.5KG
 Potassium Chromate
EW-89100-41 - POTASSIUM CHROMATE CRYSTAL ACS 500G
 Potassium Ferricyanide
EW-89100-42 - POTASSIUM FERRICYANIDE ACS 500G
 Potassium Hydroxide
EW-89100-43 - POTASSIUM HYDROXIDE PELLETS ACS 500G
EW-89100-44 - POTASSIUM HYDROXIDE PELLETS ACS 2.5KG
EW-89100-45 - POTASSIUM HYDROXIDE FLAKES Technical 2.5KG
 Potassium Iodate
EW-89100-46 - POTASSIUM IODATE POWDER ACS 100g
 Potassium Iodide
EW-89100-47 - POTASSIUM IODIDE GRANULAR ACS 500G
EW-89100-48 - POTASSIUM IODIDE GRANULAR ACS 125G
EW-89100-49 - POTASSIUM IODIDE GRANULAR USP 500G
EW-89100-50 - POTASSIUM IODIDE GRANULAR USP 2.5KG
 Potassium nitrate
EW-89100-51 - POTASSIUM NITRATE ACS 500G
 Potassium Permanganate
EW-89100-52 - POTASSIUM PERMANGANATE CRYSTAL USP 500G
 Potassium Persulfate
EW-89100-53 - POTASSIUM PERSULFATE ACS 500G
 Potassium Phosphate Monobasic
EW-89100-54 - POTASSIUM PHOSPHATE MONOBASIC ACS 2.5KG
 Propylene Glycol
EW-89100-56 - PROPYLENE GLYCOL USP 20L
 Reagent Alcohol, ACS Reagent Grade, Anhydrous, Absolute
EW-89100-57 - REAGENT ALCOHOL ABSOLUTE 500ML
EW-89100-58 - REAGENT ALCOHOL ABSOLUTE 4L
 Silver Nitrate
EW-89100-60 - SILVER NITRATE ACS 500G
EW-89100-61 - SILVER NITRATE ACS 125G
 Silver Nitrate, 0.100 Normal (N/10)
EW-89200-48 - SILVER NITRATE 0.100N 500ML
EW-89200-49 - SILVER NITRATE 0.100N 1L
EW-89200-50 - SILVER NITRATE 0.100N 4L
EW-89200-51 - SILVER NITRATE 0.100N 20L
 Silver Nitrate, 0.171 Normal, 1 mL = 10 mg NaCl
EW-89200-52 - SILVER NITRATE 0.171N 1L GLASS 1L
EW-89200-53 - SILVER NITRATE 0.171N 4L GLASS 4L
 Sodium Acetate Anhydrous
EW-89100-62 - SODIUM ACETATE ANHYDROUS ACS 2.5KG
 Sodium Bicarbonate
EW-89100-63 - SODIUM BICARBONATE ACS 500G
EW-89100-64 - SODIUM BICARBONATE ACS 2.5KG
EW-89100-65 - SODIUM BICARBONATE ACS 12KG
 Sodium Carbonate Anhydrous
EW-89100-66 - SODIUM CARBONATE ACS 500G
 Sodium Chloride
EW-89100-67 - SODIUM CHLORIDE CRYSTAL ACS 500G
EW-89100-68 - SODIUM CHLORIDE CRYSTAL ACS 2.5KG
EW-89100-69 - SODIUM CHLORIDE CRYSTAL ACS 12KG
EW-89100-70 - SOD CHLORIDE USP 500G
EW-89100-71 - SOD CHLORIDE USP 2.5KG
 Sodium fluoride
EW-89100-72 - SODIUM FLUORIDE ACS 500G
 Sodium Hydroxide
EW-89100-77 - SODIUM HYDROXIDE CRYSTAL ACS 500G
EW-89100-78 - SODIUM HYDROXIDE CRYSTAL ACS 2.5KG
EW-89100-79 - SODIUM HYDROXIDE CRYSTAL ACS 12KG
 Sodium Hydroxide Solution
EW-89100-73 - SODIUM HYDROXIDE 50% SOLUTION Reagent 500ML
EW-89100-74 - SODIUM HYDROXIDE 50% SOLUTION Reagent 1L
EW-89100-75 - SODIUM HYDROXIDE 50% SOLUTION Reagent 4L
EW-89100-76 - SODIUM HYDROXIDE 50% SOLUTION Reagent 20L
EW-89200-60 - SODIUM HYDROXIDE 0.10N 1L
EW-89200-62 - SODIUM HYDROXIDE 0.10N 4L
EW-89200-66 - SODIUM HYDROXIDE 1.0N 1L
EW-89200-67 - SODIUM HYDROXIDE 1.0N 3.8L
 Sodium Hydroxide, 0.0100 Normal (N/100)
EW-89200-54 - SODIUM HYDROXIDE 0.010N 1L
EW-89200-55 - SODIUM HYDROXIDE 0.010N 3.8L
EW-89200-56 - SODIUM HYDROXIDE 0.010N 20L
 Sodium Hydroxide, 0.0200 Normal (N/50), 1 mL = 1 mg CaCO3
EW-89200-57 - SODIUM HYDROXIDE 0.020N 1L
EW-89200-58 - SODIUM HYDROXIDE 0.020N 3.8L
 Sodium Hydroxide, 0.0500 Normal (N/20)
EW-89200-59 - SODIUM HYDROXIDE 0.050N 3.8L
 Sodium Hydroxide, 0.100 Normal (N/10)
EW-89200-61 - SODIUM HYDROXIDE 0.10N 3.8L
EW-89200-63 - SODIUM HYDROXIDE 0.10N 20L
 Sodium Hydroxide, 0.200 Normal (N/5)
EW-89200-64 - SODIUM HYDROXIDE 0.20N 1L
 Sodium Hydroxide, 0.500 Normal (N/2)
EW-89200-65 - SODIUM HYDROXIDE 0.50N 1L
 Sodium meta-Silicate Pentahydrate
EW-89100-87 - SODIUM SILICATE, META, ANHYDROUS Technical 2.5KG
 Sodium Nitrate
EW-89100-80 - SODIUM NITRATE ACS 500G
EW-89100-81 - SODIUM NITRATE ACS 2.5KG
 Sodium Phosphate Dibasic Anhydrous
EW-89100-82 - SODIUM PHOSPHATE DIBASIC ANHYDROUS ACS 500G
EW-89100-83 - SODIUM PHOSPHATE DIBASIC ANHYDROUS ACS 2.5KG
 Sodium Phosphate Dibasic Heptahydrate
EW-89100-84 - SODIUM PHOSPHATE DIBASIC HEPTA-HYDRATE ACS 500G
 Sodium phosphate, tribasic dodecahydrate,reagent ACS
EW-89100-85 - SODIUM PHOSPHATE TRIBASIC ACS 500G
 Sodium Silicate Solution
EW-89100-86 - SODIUM SILICATE 42 Be SOLUTION 4L
 Sodium sulfate
EW-89100-88 - SODIUM SULFATE ANHYDROUS ACS 2.5KG
 Sodium Thiosulfate Pentahydrate
EW-89100-89 - SODIUM THIOSULFATE PENTAHYDRATE CRYSTAL ACS 500G
 Starch Solution
EW-89200-68 - STARCH INDICATOR 0.5% (w/v) AQUEOUS 1L
EW-89200-69 - STARCH INDICATOR 0.5% (w/v) AQUEOUS 3.8L
 Sucrose
EW-89100-90 - SUCROSE ACS 500G
EW-89100-91 - SUCROSE ACS 2.5KG
 Sulfuric Acid
EW-89100-92 - SULFURIC ACID 66 Be, 93% Technical 2.5L
EW-89100-93 - SULFURIC ACID ACS 500ML
EW-89100-94 - SULFURIC ACID ACS 2.5L
EW-89100-95 - SULFURIC ACID ACS 2.5L
 Sulfuric Acid Solution
EW-89200-70 - SULFURIC ACID 0.100N 1L
EW-89200-73 - SULFURIC ACID 0.100N 4L
 Sulfuric Acid, 0.100 Normal (N/10)
EW-89200-71 - SULFURIC ACID 0.100N 1L
EW-89200-72 - SULFURIC ACID 0.100N 3.8L
EW-89200-74 - SULFURIC ACID 0.100N 20L
 Sulfuric Acid, 1.00 Normal
EW-89200-75 - SULFURIC ACID 1.00N 1L
EW-89200-76 - SULFURIC ACID 1.00N 3.8L
EW-89200-77 - SULFURIC ACID 1.00N 20L
 Sulfuric Acid, 2.00 Normal
EW-89200-78 - SULFURIC ACID 2.0N 1L
 Sulfuric Acid, 2.50 Normal
EW-89200-80 - SULFURIC ACID 2.5N 1L
EW-89200-81 - SULFURIC ACID 2.5N 3.8L
 Tetrahydrofuran
EW-89100-96 - TETRAHYDROFURAN ACS 1L
EW-89100-98 - TETRAHYDROFURAN HPLC 4L
 Toluene
EW-89100-99 - TOLUENE ACS 1L
EW-89200-01 - TOLUENE ACS 4L
 Trichloroacetic Acid
EW-89200-02 - TRICHLOROACETIC ACID ACS 500G
 Trichloroethylene
EW-89200-03 - TRICHLORETHYLENE ACS 500mL
EW-89200-04 - TRICHLORETHYLENE ACS 4L
 Universal Indicator, pH 2 - 10
EW-89200-82 - UNIVERSAL PH INDICATOR 500ML
 Xylenes
EW-89200-06 - XYLENES ACS 500mL
EW-89200-07 - XYLENES ACS 1L
EW-89200-08 - XYLENES ACS 4L
 Zinc Chloride
EW-89200-09 - ZINC CHLORIDE GRANULAR USP 2.5KG
 Zinc Sulfate Heptahydrate
EW-89200-11 - ZINC SULFATE USP 2.5KG