Cole-Parmer Technical Library
Safety Gloves Sizing Guide
Glove Sizes
Hand Circumference Method
Hand Circumference Sizing Method
Measure the circumference of your hand around the palm area with fingers together and hand relaxed. This measurement in inches is closest to your actual glove size. For example, 7" is equal to a size 7 glove. Sizes may vary between different manufacturers and styles. Except where mentioned, gloves are sized according to men's hands. For easy size comparison, see table below.
Numerical size 6-7 7-8 8-9 9-10 11
General size XS S M L XL
Length
To determine the correct glove length you need, measure from the tip of the middle finger to where the cuff edge should be on the wrist or arm; then select the proper cuff and length.
Related Links: Printable page E-mail a colleague
Complete Selection of Gloves
Safety Glove Chemical Compatibility Database
Safety Gloves Selection Guide
Safety Gloves Sizing Guide
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data