Youve heard the war stories. The
unexpected jolt that knocked
someone off his feet. The digital
multimeter that turned to toast
because it couldnt handle the
measurement it was hooked up
to perform. The guy who reached
into a tight space to adjust an
alligator clip and came out with a
little less skin on his finger.
If your job is to measure
electricity, the truth is, youre
at
risk of injury. When it comes to
testing and measuring electricity,
all that stands between you and
a nasty shock or worse is the
quality of the test equipment you
use on the job and the safety
precautions you take.
The hazards of working with
electricity arent new, of course.
But as electronic and electrical
systems become increasingly
complex, the dangers of measuring
its flow have increased and
unprepared technicians can
suddenly be faced with a simple
job gone bad.
For example, its not unheard
of for an electrician to find
himself
the victim of a transient
power spike that knocks him flat
before he knows its coming,
thanks to a big motor kicking
on or off somewhere upline.
Occasionally an electrician may
end up working with more
voltage and energy than he
realizes.
Choosing the best digital
multimeter is critical when you
work in such environments. It
can help handle an unexpected
situation, even if you wander into
dangerous territory.
The key is selecting a meter
with a high level of safety
capabilities.
Heres what to look for
next time you go shopping for a
meter:
Safety-rated fuses.
You
dont
want
a meter that uses just any
old
fuse. Specially designed
high-energy
fuses dissipate stray
voltage
before it reaches you and
are
designed to blow in time to
get
you out of harms way. Dont
make
the beginners mistake of
substituting
inferior fuses once
youve
taken the meter home,
and
dont ever disable the fuse
on
a job. Ten is a good number of
fingers
to have, and youll want
to
keep them all. Use only highenergy
fuses
approved by the
meters
manufacturer.
Component
spacing.
Proper
safety
design in a digital multimeter
begins
deep inside each
tool.
Adequate spacing between
internal
components provides a
measurable
margin of safety, but
can
be hard to check. Look for an
approval
sticker by an independent
third-party
testing laboratory.
The
lab has taken that model
apart
and stakes its reputation on
the
fact that the meter is as safe
as
its manufacturer says it is.
Independent
verification.
Dont
trust the word of just any
laboratory,
though. Look for those
whose
analysis has stood the test
of
time, such as UL, CSA, and
TUV.
Beware of wording such as
Designed
to meet specification...
Designers
plans are never a substitute
for
independent testing by
a
reputable laboratory.
Best
value, not lowest cost.
The
truth is, you often get what
you
pay for. Some cheap meters
say
theyre adequately safety
rated
and theyre not. Selecting
the
lowest-priced equipment may
be
a false economy. Treating
severe
burns from an on-the-job
electrical
accident will cost more.
Appropriate
CAT ratings.
Thats
a fancy bit of rule-making
by
the International Electrotechnical
Commission
(IEC) that sets
the
safety regulations multimeter
manufacturers
must adhere to if
their
products are marketed and
sold
in Europe. The IEC is your
friend.
The IEC has set four categories
of
electrical testing activity,
ranging
from Category I
protected
low energy circuits to
Category
IV installations where
high-powered
lines can be
exposed
to outdoor environments.
Meters
designed to the IEC
standards
will carry a category
rating
and certification label. Use
them
within their rated categories,
and
they will be more
capable
of withstanding the hazards
caused
by transients and
other
dangers in todays electrical
systems.
Always make sure your
test
tool category rating matches
how
youre using it, even if that
means
switching from meter to
meter
throughout the day. Better
yet,
invest in a good CAT IV rated
meter
and use it exclusively —
then
you never have to worry
about
which CAT level you are
working
in.
Category
II conditions are most
prevalent,
but that shouldnt lull
those
testing electrical circuits
and
sources into complacency. As
soon
as you move into an industrial
setting,
chances are youre
dealing
with Category III or Category
IV.
As soon as you get into
large
industrial motors, youre in
Category
IV territory. No matter
the
category, follow all safety
procedures
and wear appropriate
safety
equipment. And never
work
alone.
CAT-rated
accessories.
A
tester
is only as good as the
accessories
that accompany it.
Select
high-quality leads, clips
and
probes that also are
designed
with safety in mind.
Look
for finger guards and rubber
molded
grips to provide sure
touches.
Select probes slender
enough
to easily reach into tight
spots.
Most important of all, make
sure
the CAT safety ratings on
your
accessories match your
meter
— and the job at hand.
Think
of a meter and its accessories
as
a complete system. And
a
system is only as good as its
weakest
component.
Ergonomic
design. Proper
ergonomic
design makes a tester
more
than comfortable. It also
makes
it safer to use. A cushioned,
over-molded
rubber design
ensures
a strong and stable grip
and
helps prevent slipping.
So
youve selected your meter,
and
outfitted it with high-quality,
safety-rated
accessories. Youre
almost
ready to hit the job site.
Just
a few more tips to make your
safety
preparations complete.
Read
the manual! Yes,
we
know
that goes against the grain.
Do
it anyway. Manuals actually
are
chock full of valuable safety
information
and tips on making
the
most of your new equipment.
Start
at the front, the part you
usually
skip, and hang with it all
the
way through.
Replace the battery when it says to. Sure,
the manufacturer
gives
you a long lead time before
the
meter actually goes dead. But
a
failing battery is not what you
want
when all that stands
between
you and a powerful
electrical
circuit is a meter flashing,
low
battery.
Dont
ignore what you dont understand.
Theres
probably a
good
reason something doesnt
make
sense. Taking the time to
find
that reason could save your
life.
Work
safely
- Work on
de-energized circuits
whenever
possible. Use proper
lock-out/tag-out
procedures.
- When working on
live circuits,
use
protective gear: insulated
tools,
safety glasses or a face
shield,
insulated gloves.
- Stand on an
insulated mat
and
remove watches or other
jewelry.
Wear flame resistant
clothing.
- When making
measurements
on
live circuits, connect the
ground
lead first, then make
contact
with the hot lead.
Remove
the hot lead first, the
ground
lead last.
- Hang or rest
the meter if possible.
Try
to avoid holding it in
your
hands to minimize personal
exposure
to transients.
- Use the
three-point test
method
when checking to see
if
a circuit is inactive. First test
a
known live circuit. Second,
test
the target circuit. Third,
test
the live circuit again. This
verifies
that your tester
worked
properly before and
after
the measurement.
- Use the old
electricians trick
of
keeping one hand in your
pocket.
This lessens the
chance
of a closed circuit
across
your chest and through
your
heart.
|
Overvoltage
Installation
Category
|
Working
Voltage
(dc
or ac-rms to ground)
|
Peak
Impulse Transient
(20
repetitions)
|
Test
Source
(W
= V/A)
|
|
CAT
I
|
600
V
|
2500
V
|
30
Ohm source
|
|
CAT
I
|
1000
V
|
4000
V
|
30
Ohm source
|
|
CAT
II
|
600
V
|
4000
V
|
30
Ohm source
|
|
CAT
II
|
1000
V
|
6000
V
|
30
Ohm source
|
|
CAT
III
|
600
V
|
6000
V
|
30
Ohm source
|
|
CAT
III
|
1000
V
|
8000
V
|
30
Ohm source
|
|
CAT
IV
|
600
V
|
8000
V
|
30
Ohm source
|
Transient
test values for overvoltage installation categories. (50 V/150 V/300 V values
not included.)
Get
to know your CATs
Category
I — typically covers electronic equipment
- Protected
electronic equipment
- Equipment
connected to (source) circuits in
which
measures are taken to limit transient
overvoltages
to an appropriately low level
- Any
high-voltage-low-energy source derived
from
a high-winding resistance transformer,
such
as the high-voltage section of a copier.
Category
II — single-phase receptacle connected loads
- Appliance,
portable tools and other household
and
similar loads
- Outlet and long
branch circuits
- Outlets at more
than 10 meters from CAT III
source
- Outlets at more
than 20 meters from CAT IV
source
Category
III — three-phase distribution, including single-phase commercial lighting
- Equipment in
fixed installations, such as
switchgear
and polyphase motors
- Bus and feeders
in industrial plants
- Feeders and
short branch circuits, distribution
panel
devices
- Lighting
systems in larger buildings
- Appliance
outlets with short connections to
service
entrance
Category
IV — three-phase at utility connection, any outdoor conductors
- Origin of
installations, such as where lowvoltage
connection
is made to utility power
- Electricity
meters, primary overcurrent protection
equipment
- Outside and
service entrance, service drop from
pole
to building, run between meter and panel
- Overhead line
to detached building, underground
line
to well pump
|