Research in the areas of cell
biology, molecular biology, cancer, pharmaceuticals, etc. has made amazing
strides during the past decade and the technology used in those areas has had
to keep pace. The landscape (or labscape) of a typical life science laboratory
has changed dramatically over the years but the CO2 incubator
continues to be a staple in the research lab. Although the ultimate goal of
maintaining cell culture stocks has not changed, the functioning and operation
of CO2 incubators has become more accurate, more reliable and more
convenient. CO2 incubators are designed with the user and his/her
applications in mind. Three primary areas of concern when selecting a CO2
incubator are with reliability, contamination control, and ease of use. The
following overviews several of the options that are available for CO2
incubators.
CO2 Control:
The OptionsInfrared or Thermal Conductivity
CO2 levels within the chamber are
established with a set point and are controlled to maintain that set point.
When the door is opened CO2 escapes from the chamber and a CO2
sensor detects a drop in the level of CO2. CO2 is
automatically injected to raise the level to the set point. Two control
systems are available to detect changes in CO2 levelsThermal
Conductivity (TC) and Infrared (IR).
The thermal conductivity
system works by measuring the resistance between two thermistors, one exposed
to the chamber environment and the other enclosed. The presence of CO2
in the chamber changes the resistance between the two thermistors. The level
of CO2 is determined by measuring the resistance. A drawback of the
thermal conductivity system is that changes in temperature and relative
humidity can affect the accuracy of the sensor. Frequent door openings, which
cause fluctuations in temperature and relative humidity in addition to CO2
levels, can affect the accuracy of the thermal conductivity sensor.
The infrared system was
developed as an alternative to the TC sensor and is a more accurate controller
of CO2 levels. The infrared system detects CO2 levels
with an optical sensor. An air sample from the chamber is passed between an IR
emitter (a light source) and the sensor. The sensor detects a reduction in the
IR from the emitter as the CO2 in the air sample absorbs the IR.
The amount of IR absorbed is relative to the levels of CO2 in the
air sample. The IR sensor is not effected by variations in temperature and
humidity so it is more accurate than the TC sensor especially following door
openings. However, the IR system is usually more expensive than the TC system.
Temperature Control: The OptionsWater-jacketed or Radiant-walled
Maintaining a constant temperature in the incubator is
critical to the health and growth of cultured cells. There are two primary
heating options to consider when selecting a small to mid-sized CO2
incubator: water-jacketed and radiant-walled. Although both heating systems
are accurate and reliable, they each have their advantages and disadvantages.
Water-jacketed incubators maintain temperature by
surrounding the interior chamber with heated water in a separate compartment.
The water is heated and circulates around the inner chamber via natural
convection. The heat from the water radiates to the interior chamber
maintaining a constant temperature inside. Water is a particularly effective
insulator and the water-jacket system is considered a more reliable method of
heating in case of a power outage. In the wake of a power failure, a water-jacketed
incubator will hold a set temperature inside the chamber 4-5 times longer than
a radiant-walled unit
Radiant-walled incubators heat the interior chamber using
heaters mounted in the surrounding cavity that radiate heat through to the
inside chamber. A radiant-walled heating system allows for quicker recovery of
temperature following door openings or changes in temperature settings.
Radiant-walled heating systems are also more simplified for the user, not
requiring filling, monitoring, and emptying water in the water jacket.
A fan may be mounted outside of the culturing area to help
to circulate the air inside the chamber without disturbing cultures. This
gentle circulation speeds recovery of internal temperature as well as CO2
and humidity levels following door openings.
Humidity: Protecting Cultures from Desiccation
Desiccation results in lost cultures. It is important to
maintain adequate moisture inside the chamber to prevent the drying out of
cultures. Large CO2 incubators may use steam-generators or
atomizers to control relative humidity levels but most small to mid-sized
incubators use humidity pans to produce humidity through evaporation. Humidity
pans produce relative humidity levels between 95-98%. Some incubators have a
humidity reservoir that holds water in a heated pan. This increases
evaporation. A humidity reservoir may increase the relative humidity levels to
97-98% but this system is more complicated and problems may arise due to an
increase in the number of components and these units may be prone to internal
sweating.
Contamination: Exterminating the Bugs!
Contamination is a major source of frustration in cell
culturing. Manufacturers of CO2 incubators have found ways to help
combat contamination. By reducing the areas or surfaces where microorganisms
can grow and by incorporating autodecontamination cycles are ways that
manufacturers help researchers prevent contamination. Many manufacturers also
offer HEPA filters in CO2 incubators to reduce contamination during
the incubation cycle. Copper-lined chambers with copper shelves and fixtures
also reduce fungal growth and other contaminants. Removable shelves and
crevice-free interiors or coved corners in drawn internal chamber reduce the
areas where contaminants can begin growing. Surface areas are also more
accessible for the use of disinfectants. Some CO2 incubators also
have autodecontamination cycles that decontaminate the internal chamber in
between incubation cycles. Autodecontamination works by raising the internal
temperature to +90oC for several hours thereby killing contaminating
microorganisms. The autodecontamination cycle, used in conjunction with HEPA
filters, greatly reduces contamination.
Ease of Use
A CO2 incubator should be easy to use and maintain.
With the introduction of microprocessor controls and various accessories, the
CO2 incubator is approaching a set it and forget it mode of
operation. Features such as over-temperature thermostats and alarms, CO2
alarms, door opening alarms, password protection of settings, self-calibration,
and autodecontamination cycles offer easy operation and security for the user.
Other features such as stackable models help conserve valuable lab space. Some
models require stacking kits sold as accessories while other units are
stackable without kits or tools. Some models also offer exchangeable door
swings to accommodate lab space and placement.
Selecting the size of incubator to meet your storage needs
and space limitations contributes to the ease of use. CO2
incubators range in size from personal bench-top incubators (<40 liters)
to large-capacity incubators (>700 liters). The mid-sized models (140-180
liters) offer the most options described above but the smaller and larger
capacity incubators accommodate specific storage requirements. The broad range
of sizes, options, and accessories for CO2 incubators are quite
numerous and should meet any researchers needs.