A new fritted filter should be washed by suction with hot hydrochloric acid and then rinsed with water before it is used. This treatment will remove loose particles of foreign matter such as dust. It is advisable to clean all Pyrex® brand fritted filters as soon as possible after use. This will prolong their life.
Many precipitates can be removed from the filter surface simply by rinsing from the reverse side with water under pressure not exceeding 15 lbs/square inch. Drawing water through the filter from the reverse side with a vacuum pump is also effective. Some precipitates tend to clog the pores of a fritted filter. Treatment here must be by chemical means.
A few suggestions found generally useful follow:
| Material | Cleaning solution |
| Fatty materials | Carbon tetrachloride |
| Organic matter* | Hot concentrated cleaning solution, or hot concentrated sulfuric acid plus a few drops of sodium or potassium nitrite |
| Albumen | Hot ammonia or hot hydrochloric acid |
| Glucose | Hot mixed acid: H2SO4 + HNO3 |
Copper or iron oxides | Hot hydrochloric acid plus potassium chlorate |
| Mercury residue | Hot nitric acid |
| Silver chloride | Ammonia or sodium hyposulfite |
| Viscose | 5 to 10% NaOH, followed by cleaning solution |
| Aluminous and siliceous residues | 2% hydrofluoric acid followed by concentrated sulfuric acid; rinse immediately with distilled water followed by a few mL of acetone. Repeat rinsing until all traces of acid are removed. |
*Cleaning solution containing bichromate will tend to permanently stain fritted ware. This is undesirable for biochemical or pharmaceutical work, and hence the sulfuric-nitrite solution is suggested.
Go to Top
The resistance to thermal shock of fritted ware is less than that of non-porous Pyrex brand labware. Therefore, articles of fritted ware should not be subjected to excessive temperature changes or to direct flame.
Dry fritted crucibles at room temperature may be placed into a drying oven operating at 150°C (302°F).
Fritted ware may be safely heated in a furnace to 500°C (932°F) without ill effect, provided that the cycle of heating and cooling is gradual.
Go to Top