Cole-Parmer Technical Library
Effects of Chemicals on Labware
Chemicals may affect the weight, strength, color, dimensions, flexibility, and surface appearance of labware. The basic models of interaction that cause these changes are: (1) chemical attack on the polymer chain, with resultant reduction in physical properties, including oxidation; reaction of functional groups in or on the chain; and depolymerization; (2) physical change, including absorption of solvents, resulting in softening and swelling of the plastic; permeation of solvent through the plastic; or dissolution in a solvent; and (3) stress-cracking from the interaction of a "stress-cracking agent" with molded-in or external stresses. The reactive combination of compounds of two or more classes may cause a synergistic or undesirable chemical effect. Other factors affecting chemical resistance include: temperature, pressure, internal or external stresses (such as centrifugation), and length of exposure to and concentration of the chemical. As temperature increases, resistance to attack decreases.
Related Links: Printable page E-mail a colleague
Complete Selection of Labware
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data