Cole-Parmer Technical Library
Cleanroom Garments
Cleanroom Gowning Sequence
The correct order for cleanroom gowning:
  1. Shoe covers
  2. Cap
  3. Hood
  4. Face mask
  5. Coveralls
  6. Boots
  7. Safety glasses
  8. Gloves
Garment Design
It is very important that garments be sized correctly. Improperly sized garments can raise the level of contaminants and cause safety concerns.
Cleanroom garments must have these basic characteristics:
  • A minimum of seams. The seams must firmly envelop the edge of the material.
  • Be loose fitting to eliminate abrasion
  • Absence of pockets, belts, and pleated or tucked areas.
  • Tight fabric weave to minimize the passage of dust.
  • Fabric must reduce static electricity generation and still remain nonlinting.
  • Garments should cover as much of the person as possible, especially around the neck area.
Related Links: Printable page E-mail a colleague
Complete Selection of Apparel
Complete Selection of Gloves
Cleanrooms
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data