Cole-Parmer Technical Library
Conductivity Values µS/cm at 77°F (25°C)
% Weight ppm
mg/l
NaCl NaOH HCl Acetic
acid
0.0001 1 2.2 6.2 11.7 4.2
0.0003 3 6.5 18.4 35.0 7.4
0.001 10 21.4 61.1 116 15.5
0.003 30 64 182 340 30.6
0.01 100 210 603 1140 63
0.03 300 617 1780 3390 114
0.1 1000 1990 5820 11,100 209
0.3 3000 5690 16,900 32,200 368
1.0 10,000 17,600 53,200 103,000 640
3.0 48,600 144,000 283,000 1120
5.0 78,300 223,000 432,000 1230
10.0 140,000 358,000 709,000 1530
20.0 226,000 414,000 850,000 1600
30.0 Saturated 292,000 732,000 1405
40.0 Saturated 191,000 Saturated 1080
50.0 Saturated 150,000 Saturated 740
75.0 Saturated Saturated Saturated 168
100.0 Saturated Saturated Saturated <1
Go to Top
Related Links: Printable page E-mail a colleague
Complete Selection of Conductivity Controllers
Complete Selection of Conductivity/Resistivity/TDS Meters
Conductivity FAQs
Conductivity Technical Information
Conductivity Theory and Technical Tips
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data