Cole-Parmer Technical Library
Air Consumption of a Cylinder
To calculate the air consumption of a cylinder, multiply the total inches of stroke (extend plus retract) by the cycles per minute times the SCFM factor from the chart below.
Air consumption = Stroke length (in.) x Cycles/min x SCFM factor
To find the SCFM factor, find your gauge pressure in the left hand column. Next, find your cylinder bore size in the chart. Where the two intersect, read down to the SCFM factor at the bottom of the chart.
Air Consumption Chart
Related Links: Printable page E-mail a colleague
Complete Selection of Actuators
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data