Electrical Data
Electrical Terminology
Electrical Formulas
Properties of Conductors Multiplying Factors for Converting Resistance to 60-Hertz AC Resistance
Volt (E) is the unit of electric pressure or electromotive force. It is the potential that will produce a current of 1 ampere through a resistance of 1 ohm.
Ampere (I) is the unit of electrical current (coulombs per second).
Ohm (R) is the unit of electrical resistance (volts/ampere).
Watts (W) and kilowatts (KW) are units of electric power.
Kilovolt-ampere (KVA) is a measurement of apparent electric power.
Kilowatt hour (Kwhr) is a unit of electrical energy or work performed.
Joule (J) is a metric unit of energy: watt per second.
1 Kwhr = 2,655,000 ft-lb = 1.341 hp-hr = 3413 Btu = 3,600,000 joules.
Ohm law relationships (direct current)
E = IR + W/I = √WR
W = I2R = E2/R = EI
I = E/R = W/E = √W/R
R = E/I = W/I2 = E2/W
For electrical formulas, see the chart below. Use the symbols above plus the ones defined in the text above the table.
Go to Top
Use the symbols in Electrical Terminology above, plus:
eff = efficiency (expressed as a decimal)
pf = power factor (expressed as a decimal)
hp = horsepower output
| Required | Direct current | Alternating current |
| Single-phase | 3-phase* |
| KVA | | IE 1000 | 1.73 IE 1000 |
| Kilowatts | IE 1000 | IE (pf) 1000 | 1.73 IE (pf) 1000 |
| Horsepower (output) | IE (eff)/746 | IE (eff)(pf)/746 | 1.73 IE (eff)(pf)/746 |
| Joules | IE/sec | IE (eff)(pf)/sec | 1.73 IE (eff)(pf)/sec |
| Amperes (hp known) | 746 (hp)/E (eff) | 746 (hp)/E (eff)(pf) | 746 (hp)/1.73 E (eff)(pf) |
| Amperes (KW known) | 1000 KW/E | 1000 KW/E (pf) | 1000 KW/1.73 E (pf) |
| Amperes (kva known) | | 1000 KVA/E | 1000 KVA/1.73 E |
*For 3-phase systems, E is measured line to line and I is phase current.
Go to Top
Size (AWG MCM) | Area circle (mils) | Concentric lay conductors | Bare conductors | DC resistance Ω/M ft at 77°F (25°C)** |
No. of wires | Diameter each wire (inch) | Diameter (inch) | Area (sq inch) | Copper | Aluminum |
| Bare | Tin'd |
| 18 | 1620 | | | 0.0403 | 0.0013 | 6.51 | 6.79 | 10.7 |
| 16 | 2580 | | | 0.0508 | 0.0020 | 4.10 | 4.26 | 6.72 |
| 14 | 4110 | | | 0.0641 | 0.0032 | 2.57 | 2.68 | 4.22 |
| 12 | 6530 | | | 0.0808 | 0.0051 | 1.62 | 1.68 | 2.66 |
| 10 | 10,380 | | | 0.1019 | 0.0081 | 1.018 | 1.06 | 1.67 |
| 8 | 16,510 | | | 0.1285 | 0.0130 | 0.6404 | 0.659 | 1.05 |
| 6 | 26,240 | 7 | 0.0612 | 0.184 | 0.027 | 0.410 | 0.427 | 0.674 |
| 4 | 41,740 | 7 | 0.0772 | 0.232 | 0.042 | 0.259 | 0.269 | 0.424 |
| 3 | 52,620 | 7 | 0.0867 | 0.260 | 0.053 | 0.205 | 0.213 | 0.336 |
| 2 | 66,360 | 7 | 0.0974 | 0.292 | 0.067 | 0.162 | 0.169 | 0.266 |
| 1 | 83,690 | 19 | 0.0664 | 0.332 | 0.087 | 0.129 | 0.134 | 0.211 |
| 0 | 105,600 | 19 | 0.0745 | 0.372 | 0.109 | 0.102 | 0.106 | 0.168 |
| 00 | 133,100 | 19 | 0.0837 | 0.418 | 0.137 | 0.0811 | 0.0843 | 0.133 |
| 000 | 167,800 | 19 | 0.0940 | 0.470 | 0.173 | 0.0642 | 0.0668 | 0.105 |
| 0000 | 211,600 | 19 | 0.1055 | 0.528 | 0.219 | 0.0509 | 0.0525 | 0.0836 |
| 250 | 250,000 | 37 | 0.0822 | 0.575 | 0.260 | 0.0431 | 0.0449 | 0.0708 |
| 300 | 300,000 | 37 | 0.0900 | 0.630 | 0.312 | 0.0360 | 0.0374 | 0.0590 |
| 350 | 350,000 | 37 | 0.0973 | 0.681 | 0.364 | 0.0308 | 0.0320 | 0.0505 |
| 400 | 400,000 | 37 | 0.1040 | 0.728 | 0.416 | 0.0270 | 0.0278 | 0.0442 |
| 500 | 500,000 | 37 | 0.1162 | 0.813 | 0.519 | 0.0216 | 0.0222 | 0.0354 |
| 600 | 600,000 | 61 | 0.0992 | 0.893 | 0.626 | 0.0180 | 0.0187 | 0.0295 |
| 700 | 700,000 | 61 | 0.1071 | 0.964 | 0.730 | 0.0154 | 0.0159 | 0.0253 |
| 750 | 750,000 | 61 | 0.1109 | 0.998 | 0.782 | 0.0144 | 0.0148 | 0.0236 |
| 800 | 800,000 | 61 | 0.1145 | 1.030 | 0.833 | 0.0135 | 0.0139 | 0.0221 |
| 900 | 900,000 | 61 | 0.1215 | 1.090 | 0.933 | 0.0120 | 0.0123 | 0.0197 |
| 1000 | 1,000,000 | 61 | 0.1280 | 1.150 | 1.039 | 0.0108 | 0.0111 | 0.0177 |
| 1250 | 1,250,000 | 91 | 0.1172 | 1.289 | 1.305 | 0.00863 | 0.00888 | 0.0142 |
| 1500 | 1,500,000 | 91 | 0.1284 | 1.140 | 1.561 | 0.00719 | 0.00740 | 0.0118 |
| 1750 | 1,750,000 | 127 | 0.1174 | 1.526 | 1.829 | 0.00616 | 0.00634 | 0.0101 |
| 2000 | 2,000,000 | 127 | 0.1255 | 1.630 | 2.087 | 0.00539 | 0.00555 | 0.00885 |
** The resistance values given in the last three columns apply only to direct current. When using conductors larger than number 4/0 with alternating current, use the multiplying factors in the table below to compensate for skin effect.
Go to Top
| Size | Multiplying factor |
For nonmetallic sheathed cables in air or nonmetallic conduits | For metallic sheathed cables or all cables in metallic raceways |
| Copper | Aluminum | Copper | Aluminum |
| Up to 3 AWG | 1.000 | 1.000 | 1.00 | 1.00 |
| Up to 2 AWG | 1.000 | 1.000 | 1.01 | 1.00 |
| Up to 1 AWG | 1.000 | 1.000 | 1.01 | 1.00 |
| 0 | 1.001 | 1.000 | 1.02 | 1.00 |
| 00 | 1.001 | 1.001 | 1.03 | 1.00 |
| 000 | 1.002 | 1.001 | 1.04 | 1.01 |
| 0000 | 1.004 | 1.002 | 1.05 | 1.01 |
| 250 MCM | 1.005 | 1.002 | 1.06 | 1.02 |
| 300 MCM | 1.006 | 1.003 | 1.07 | 1.02 |
| 350 MCM | 1.009 | 1.004 | 1.08 | 1.03 |
| 400 MCM | 1.011 | 1.005 | 1.10 | 1.04 |
| 500 MCM | 1.018 | 1.007 | 1.13 | 1.06 |
| 600 MCM | 1.025 | 1.010 | 1.16 | 1.08 |
| 700 MCM | 1.034 | 1.013 | 1.19 | 1.11 |
| 750 MCM | 1.039 | 1.015 | 1.21 | 1.12 |
| 800 MCM | 1.044 | 1.017 | 1.22 | 1.14 |
| 1000 MCM | 1.067 | 1.026 | 1.30 | 1.19 |
| 1250 MCM | 1.102 | 1.040 | 1.41 | 1.27 |
| 1500 MCM | 1.142 | 1.058 | 1.53 | 1.36 |
| 1750 MCM | 1.185 | 1.079 | 1.67 | 1.46 |
| 2000 MCM | 1.233 | 1.100 | 1.82 | 1.56 |
Go to Top
Complete Selection of Electrical/Electronic Test Equipment