Emissivity of Specific Materials
The emissivity information below was determined under ideal conditions. Note: Surface contamination such as dust, oil films, or other agents will affect the actual emissivity of your material.
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Adobe | 68 | 20 | 0.90 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Alloys: |
| Alloy 24ST, polished | 75 | 24 | 0.90 |
| Alloy 75ST | 75 | 24 | 0.11 |
| 20-Ni, 25-Cr, 55-Fe, oxidized | 392 932 | 200 500 | 0.90 0.97 |
| 60-Ni, 12-Cr, 28-Fe, oxidized | 518 1040 | 270 560 | 0.89 0.82 |
| 80-Ni, 20-Cr, oxidized | 212 1112 2372 | 100 600 1300 | 0.87 0.87 0.89 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Aluminum: |
| Unoxidized | 77 212 932 | 25 100 500 | 0.02 0.03 0.06 |
| Oxidized | 390 1110 | 199 599 | 0.11 0.19 |
| Oxidized @1110°F | 390 1110 | 199 599 | 0.11 0.19 |
| Heavily oxidized | 200 940 | 93 504 | 0.20 0.31 |
| Highly polished | 212 | 100 | 0.09 |
| Roughly polished | 212 | 100 | 0.18 |
| Commercial sheet | 212 | 100 | 0.09 |
| Highly polished plate | 440 1070 | 227 577 | 0.04 0.06 |
| Bright rolled plate | 338 932 | 170 500 | 0.04 0.05 |
| Alloy A3003, oxidized | 600 900 | 315 482 | 0.40 0.40 |
| Alloy 1100-0 | 200 to 800 | 93 to 427 | 0.05 |
| Alloy 24ST | 75 | 24 | 0.09 |
| Alloy 24ST, polished | 75 | 24 | 0.09 |
| Alloy 75ST | 75 | 24 | 0.11 |
| Alloy 75ST, polished | 75 | 24 | 0.08 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Asphalt: |
| Pavement | 100 | 38 | 0.93 |
| Tar paper | 68 | 20 | 0.93 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Basalt | 68 | 20 | 0.72 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Bismuth: |
| Bright | 176 | 80 | 0.34 |
| Unoxidized | 77 212 | 25 100 | 0.05 0.06 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Brass: |
| Matte | 68 | 20 | 0.07 |
| Burnished to brown color | 68 | 20 | 0.40 |
| Cu-Zn, brass oxidized | 392 752 1112 | 200 400 600 | 0.61 0.60 0.61 |
| Unoxidized | 77 | 25 | 0.04 |
| Unoxidized | 212 | 100 | 0.04 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Brick: |
| Red, rough | 70 | 21 | 0.93 |
| Gault cream | 2500 to 5000 | 1371 to 2760 | 0.26 to 0.30 |
| Fire clay | 2500 | 1371 | 0.75 |
| Light buff | 1000 | 538 | 0.80 |
| Lime clay | 2500 | 1371 | 0.43 |
| Fire brick | 1832 | 1000 | 0.75 to 0.80 |
| Magnesite, refractory | 1832 | 1000 | 0.38 |
| Gray brick | 2012 | 1100 | 0.75 |
| Silica, glazed | 2000 | 1093 | 0.88 |
| Silica, unglazed | 2000 | 1093 | 0.80 |
| Sandime | 2500 to 5000 | 1371 to 2760 | 0.59 to 0.63 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Cadmium | 77 | 25 | 0.02 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Carbon: |
| Lamp black | 77 | 25 | 0.95 |
| Unoxidized | 77 | 25 | 0.81 |
| Unoxidized | 212 932 | 100 500 | 0.81 0.79 |
| Candle soot | 250 | 121 | 0.95 |
| Filament | 500 | 260 | 0.95 |
| Graphitized | 212 572 932 | 100 300 500 | 0.76 0.75 0.71 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Carborundum | 1850 | 1010 | 0.92 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Cast iron: |
| Oxidized | 390 1110 | 199 599 | 0.64 0.78 |
| Unoxidized | 212 | 100 | 0.21 |
| Strong oxidation | 104 482 | 40 250 | 0.95 0.95 |
| Liquid | 2795 | 1535 | 0.29 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Ceramic: |
| Alumina on Inconel | 800 to 2000 | 427 to 1093 | 0.69 to 0.45 |
| Earthenware, glazed | 70 | 21 | 0.90 |
| Earthenware, matte | 70 | 21 | 0.93 |
| Greens, No. 5210-2C | 200 to 750 | 93 to 399 | 0.89 to 0.82 |
| Coating, No. C20A | 200 to 750 | 93 to 399 | 0.73 to 0.67 |
| Porcelain | 72 | 22 | 0.92 |
| White, Al2O3 | 200 | 93 | 0.90 |
| Zirconia on Inconel | 800 to 2000 | 427 to 1093 | 0.62 to 0.45 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Chromium: |
| Chromium | 100 1000 | 38 538 | 0.08 0.26 |
| Chromium, polished | 302 | 150 | 0.06 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Clay: | 68 | 20 | 0.39 |
| Fired | 158 | 70 | 0.91 |
| Shale | 68 | 20 | 0.69 |
| Tiles, light red | 2500 to 5000 | 1371 to 2760 | 0.32 to 0.34 |
| Tiles, red | 2500 to 5000 | 1371 to 2760 | 0.40 to 0.51 |
| Tiles, dark purple | 2500 to 5000 | 1371 to 2760 | 0.78 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Columbium: |
| Unoxidized | 1500 2000 | 816 1093 | 0.19 0.24 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Concrete: |
| Rough | 32 to 200 | 0 to 93 | 0.94 |
| Tiles, natural | 2500 to 5000 | 1371 to 2760 | 0.63 to 0.62 |
| Tiles, brown | 2500 to 5000 | 1371 to 2760 | 0.87 to 0.83 |
| Tiles, black | 2500 to 5000 | 1371 to 2760 | 0.94 to 0.91 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Copper: |
| Black, oxidized | 100 | 38 | 0.78 |
| Etched | 100 | 38 | 0.09 |
| Matte | 100 | 38 | 0.22 |
| Roughly polished | 100 | 38 | 0.07 |
| Polished | 100 | 38 | 0.03 |
| Highly polished | 100 | 38 | 0.02 |
| Rolled | 100 | 38 | 0.64 |
| Rough | 100 | 38 | 0.74 |
| Molten | 1000 1970 2230 | 538 108 1221 | 0.15 0.16 0.13 |
| Nickel plated | 100 to 500 | 38 to 260 | 0.37 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Cotton cloth | 68 | 20 | 0.77 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Dolomite lime | 68 | 20 | 0.41 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Dow metal | 0 to 600 | -18 to 315 | 0.15 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Emery corundum | 176 | 80 | 0.86 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Glass: |
| Convex D | 212 600 932 | 100 315 500 | 0.80 0.80 0.76 |
| Nonex | 212 600 932 | 100 315 500 | 0.82 0.82 0.78 |
| Smooth | 32 to 200 | 0 to 93 | 0.92 to 0.94 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Gold: |
| Enamel | 212 | 100 | 0.37 |
| Plate (0.001) on 0.0005 silver | 200 to 750 | 93 to 399 | 0.11 to 0.14 |
| Plate (0.001) on 0.005 nickel | 200 to 750 | 93 to 399 | 0.07 to 0.09 |
| Polished | 100 to 500 1000 to 2000 | 38 to 260 538 to 1093 | 0.02 0.03 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Granite | 70 | 21 | 0.45 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Gravel | 100 | 38 | 0.28 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Gypsum | 68 | 20 | 0.80 to 0.90 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Haynes Alloy C., oxidized | 600 to 2000 | 316 to 1093 | 0.90 to 0.96 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Haynes Alloy 25, oxidized | 600 to 2000 | 316 to 1093 | 0.86 to 0.89 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Haynes Alloy X., oxidized | 600 to 2000 | 316 to 1093 | 0.85 to 0.88 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Ice: |
| Smooth | 32 | 0 | 0.97 |
| Rough | 32 | 0 | 0.98 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Inconel: |
| Sheet | 1000 1200 1400 | 538 649 760 | 0.28 0.42 0.58 |
| Inconel X, polished | 75 | 24 | 0.19 |
| Inconel B, polished | 75 | 24 | 0.21 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Iron: |
| Oxidized | 212 930 2190 | 100 499 1199 | 0.74 0.89 0.89 |
| Unoxidized | 212 | 100 | 0.05 |
| Red rust | 77 | 25 | 0.70 |
| Rusted | 77 | 25 | 0.65 |
| Liquid | 2760 to 3220 | 1516 to 1771 | 0.42 to 0.45 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Lacquer: |
| Black | 200 | 93 | 0.96 |
| Blue, on aluminum foil | 100 | 38 | 0.78 |
| Clear, on aluminum foil, 2 coats | 200 | 93 | 0.08 (0.09) |
| Clear, on bright copper | 200 | 93 | 0.66 |
| Clear, on tarnished copper | 200 | 93 | 0.64 |
| Red, on aluminum foil, 2 coats | 100 | 38 | 0.61 (0.74) |
| White, on aluminum foil, 2 coats | 100 | 38 | 0.69 (0.88) |
| White | 200 | 93 | 0.95 |
| Yellow, on aluminum foil, 2 coats | 100 | 38 | 0.57 (0.79) |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Lead: |
| Polished | 100 to 500 | 38 to 260 | 0.06 to 0.08 |
| Rough | 100 | 38 | 0.43 |
| Oxidized | 100 | 38 | 0.43 |
| Oxidized @ 1100¡F | 100 | 38 | 0.63 |
| Oxidized, gray | 100 | 38 | 0.28 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Lime mortar | 100 to 500 | 38 to 260 | 0.90 to 0.92 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Limestone | 100 | 38 | 0.95 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Magnesium | 100 to 500 | 38 to 260 | 0.07 to 0.13 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Magnesium oxide | 1800 to 3140 | 982 to 1727 | 0.16 to 0.20 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Marble: |
| White | 100 | 38 | 0.95 |
| Smooth | 100 | 38 | 0.56 |
| Polished gray | 100 | 38 | 0.75 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Mercury | 32 77 100 212 | 0 25 38 100 | 0.09 0.10 0.10 0.12 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Mica | 100 | 38 | 0.75 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Oil on nickel: |
| 0.001 film | 72 | 22 | 0.27 |
| 0.002 film | 72 | 22 | 0.46 |
| 0.005 film | 72 | 22 | 0.72 |
| Thick film | 72 | 22 | 0.82 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Oil, linseed: |
| On aluminum foil, uncoated | 250 | 121 | 0.09 |
| On aluminum foil, 1 coat | 250 | 121 | 0.56 |
| On aluminum foil, 2 coats | 250 | 121 | 0.51 |
| On polished iron, 0.001 film | 100 | 38 | 0.22 |
| On polished iron, 0.002 film | 100 | 38 | 0.45 |
| On polished iron, 0.004 film | 100 | 38 | 0.65 |
| On polished iron, thick film | 100 | 38 | 0.83 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Paints: |
| Blue, CO2O3 | 75 | 24 | 0.94 |
| Black, CuO | 75 | 24 | 0.96 |
| Green, Cu2O3 | 75 | 24 | 0.92 |
| Red, Fe2O3 | 75 | 24 | 0.91 |
| White, Al2O3 | 75 | 24 | 0.94 |
| White, Y2O3 | 75 | 24 | 0.90 |
| White, ZnO | 75 | 24 | 0.95 |
| White, MgCO3 | 75 | 24 | 0.91 |
| White, ZrO2 | 75 | 24 | 0.95 |
| White, ThCO2 | 75 | 24 | 0.90 |
| White, MgO | 75 | 24 | 0.91 |
| White, PbCO3 | 75 | 24 | 0.93 |
| Yellow, PbO | 75 | 24 | 0.90 |
| Yellow, PbCrO4 | 75 | 24 | 0.93 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Paints, aluminum: | 100 | 38 | 0.27 to 0.67 |
| 10% Al | 100 | 38 | 0.52 |
| 26% Al | 100 | 38 | 0.30 |
| DOW XP-310 | 200 | 93 | 0.22 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Paints, bronze: | Low | 0.34 to 0.80 |
| Gum varnish, 2 coats | 70 | 21 | 0.53 |
| Gum varnish, 3 coats | 70 | 21 | 0.50 |
| Cellulose binder, 2 coats | 70 | 21 | 0.34 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Paints, oil: |
| All colors | 200 | 93 | 0.92 to 0.96 |
| Black | 200 | 93 | 0.92 |
| Black gloss | 70 | 21 | 0.90 |
| Camouflage green | 125 | 52 | 0.85 |
| Flat black | 80 | 27 | 0.88 |
| Flat white | 80 | 27 | 0.91 |
| Gray green | 70 | 21 | 0.95 |
| Green | 200 | 93 | 0.95 |
| Lamp black | 209 | 98 | 0.96 |
| Red | 200 | 93 | 0.95 |
| White | 200 | 93 | 0.94 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
Palladium plate (0.00005) on 0.0005 silver | 200 to 750 | 93 to 399 | 0.16 to 0.17 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Paper: |
| Kraft (average non-glass) | | | 0.90 |
| Offset (glass) | | | 0.50 to 0.60 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Plastics (average) | Low | 0.95 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Platinum: |
| Platinum, black | 100 500 2000 | 38 260 1093 | 0.93 0.96 0.97 |
| Platinum, oxidized @ 1100°F | 500 1000 | 260 538 | 0.07 0.11 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Quartz: |
| Rough, fused: | 70 | 21 | 0.93 |
| Glass, 1.98 mm | 540 1540 | 282 838 | 0.90 0.41 |
| Glass, 6.88 mm | 540 1540 | 282 838 | 0.93 0.47 |
| Opaque | 570 1540 | 299 838 | 0.92 0.68 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Red lead | 212 | 100 | 0.93 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
Rhodium flash: 0.0002 on 0.0005 Ni | 200 to 700 | 93 to 271 | 0.10 to 0.18 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Rubber: |
| Hard | 74 | 23 | 0.94 |
| Soft, gray | 76 | 24 | 0.86 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Sand | 68 | 20 | 0.76 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Sawdust | 68 | 20 | 0.75 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Silica: |
| Glazed | 1832 | 1000 | 0.85 |
| Unglazed | 2012 | 1100 | 0.75 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Silver: |
| Plate 0.0005 on nickel | 200 to 700 | 93 to 271 | 0.06 to 0.07 |
| Polished | 100 500 1000 2000 | 38 260 538 1093 | 0.01 0.02 0.03 0.03 |
| Slate | 100 | 38 | 0.67 to 0.80 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Snow: |
| Fine particles | 20 | 7 | 0.82 |
| Granular | 18 | 8 | 0.89 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Soil: |
| Surface | 100 | 38 | 0.38 |
| Black loam | 68 | 20 | 0.66 |
| Plowed field | 68 | 20 | 0.38 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Steel: |
| Cold rolled | 200 | 93 | 0.75 to 0.85 |
| Ground sheet | 1720 to 2010 | 938 to 1099 | 0.55 to 0.61 |
| Polished sheet | 100 500 1000 | 38 260 538 | 0.07 0.10 0.14 |
| Mild steel, polished | 75 | 24 | 0.10 |
| Mild steel, smooth | 75 | 24 | 0.12 |
| Mile steel, liquid | 2910 to 3270 | 1599 to 1799 | 0.28 |
| Unoxidized | 77 212 | 25 100 | 0.80 0.08 |
| Oxidized | 77 | 25 | 0.80 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Steel alloys: |
| Type 301, polished | 75 450 1740 | 24 232 949 | 0.27 0.57 0.55 |
| Type 303, oxidized | 600 to 2000 | 316 to 1093 | 0.74 to 0.87 |
| Type 310, rolled | 1500 to 2100 | 816 to 1149 | 0.56 to 0.81 |
| Type 316, polished | 75 450 1740 | 24 232 949 | 0.28 0.57 0.66 |
| Type 321 | 200 to 800 | 93 to 427 | 0.27 to 0.32 |
| Type 321, polished | 300 to 1500 | 149 to 816 | 0.18 to 0.49 |
| Type 321, with black oxide | 200 to 800 | 93 to 427 | 0.66 to 0.76 |
| Type 347, oxidized | 600 to 2000 | 316 to 1093 | 0.87 to 0.91 |
| Type 350 | 200 to 800 | 93 to 427 | 0.18 to 0.27 |
| Type 350, polished | 300 to 1800 | 149 to 982 | 0.11 to 0.35 |
| Type 446, polished | 300 to 1500 | 419 to 816 | 0.15 to 0.37 |
| Type 17-7PH | 200 to 600 | 93 to 315 | 0.44 to 0.51 |
| Type 17-7PH, polished | 300 to 1500 | 149 to 816 | 0.09 to 0.16 |
| Type C1020, oxidized | 600 to 2000 | 316 to 1093 | 0.87 to 0.91 |
| Type PH-15-7MO | 300 to 1200 | 149 to 649 | 0.07 to 0.19 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Stellite, polished | 68 | 20 | 0.18 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Stonework | 100 | 38 | 0.93 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Tantalum: |
| Unoxidized | 1340 2000 3600 5306 | 727 1093 1982 2930 | 0.14 0.19 0.26 0.30 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Tin: |
| Unoxidized | 77 212 | 25 100 | 0.04 0.05 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Tinned iron: |
| Bright | 76 212 | 24 100 | 0.05 0.08 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Titanium: |
| Alloy C110M, polished | 300 to 1200 | 149 to 649 | 0.08 to 0.19 |
| Alloy C110M, oxidized @ 1000°F | 200 to 800 | 93 to 427 | 0.51 to 0.61 |
| Alloy TI-95A, oxidized @ 1000°F | 200 to 800 | 93 to 427 | 0.35 to 0.48 |
| Anodized onto stainless steel | 200 to 600 | 93 to 316 | 0.96 to 0.82 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Tungsten: |
| Unoxidized | 77 212 932 1832 2732 3632 | 25 100 500 1000 1500 2000 | 0.02 0.03 0.07 0.15 0.23 0.28 |
| Filament (aged) | 100 1000 5000 | 38 538 2760 | 0.03 0.11 0.35 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Uranium oxide | 1880 | 1027 | 0.79 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Water | 100 | 38 | 0.93 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Wood: | Low | 0.80 to 0.90 |
| Beech, planed | 158 | 70 | 0.94 |
| Oak, planed | 100 | 38 | 0.91 |
| Spruce, sanded | 100 | 38 | 0.89 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Wrought iron: |
| Dull | 77 660 | 25 349 | 0.94 0.94 |
| Smooth | 100 | 38 | 0.35 |
| Polished | 100 | 38 | 0.28 |
| Go to Top
|
| MATERIAL | TEMPERATURE | EMISSIVITY |
| °F | °C |
| Zinc: |
| Bright galvanized | 100 | 38 | 0.23 |
| Commercial 99.1% | 500 | 260 | 0.05 |
| Galvanized | 100 | 38 | 0.28 |
| Oxidized | 500 to 1000 | 260 to 538 | 0.11 |
| Polished | 100 500 1000 2000 | 38 260 538 1093 | 0.02 0.03 0.04 0.06 |
Go to Top
Complete Selection of Infrared Thermometers
Advancements in Temperature: Infrared Technology
Infrared Thermometers FAQs
Temperature Conversion Equations
Temperature FAQs
Temperature Instrument Ranges
When to Use A Two-Color Infrared Thermometer