  Cart  Subtotal = 0.00
1-888-358-4717 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Cole-Parmer Technical Library
Fluid Fundamentals
Fluids include liquids, gases, and mixtures of liquids, solids, and gases. For the purposes of this reference book, the terms fluid and liquid are both used to mean a pure liquid or a liquid mixed with gases or solids that acts essentially like a liquid in a pumping application.
Density or specific weight of a fluid is its weight per unit volume, often expressed in units of grams per cubic centimeter or pounds per cubic foot. The density of a fluid changes with temperature.
Example:
If weight is 36.36 kg (80 lb), density is 1.28 g/cm3 (80 lb/cu ft).
Specific gravity of a fluid is the ratio of its density to the density of water. Since it is a ratio, it has no units associated with it.
Example:
Specific gravity = 80 lb/62.4 lb or, Specific gravity = 1.282
Related Links: Printable page E-mail a colleague
General Pump FAQs
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Terminology
Liquid Pumps Technical Equations and Information Tables
Pump Types and Definitions
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data