Cole-Parmer Technical Library
Installing Your Paddle-Wheel Flow Sensor
For best results, allow a straight run of pipe before and after the sensor after any bends, valves, or flow restrictions.
Stated accuracy is not guaranteed unless the Signet installation fittings are used. The installation fitting ensures proper paddle depth and orientation.
In horizontal pipe runs with no air pockets or sediments present, mount the sensor/fitting in the 12 o'clock or 6 o'clock position. If sediment or air pockets are present, tilt the sensor/fitting at a maximum angle of 45° to avoid these obstacles. Vertical runs require upward flow. Pipes must be full.
Related Links: Printable page E-mail a colleague
Complete Selection of Flowmeters
Flowmeter Applications
How Volumetric Flowmeters Work
Installing Your Paddle-Wheel Flow Sensor
Selecting the Right Flowmeter—Part 1
Selecting the Right Flowmeter—Part 2
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data