USB provides test engineers with an inexpensive, easy-to-use, high-speed bus-integration
standard for computer-based measurement systems. The standard, however, does not mandate
signal-circuit isolation. This leaves those who would use USB for test and measurement
applications open to a number of gotchas. To avoid problems, engineers need to understand
isolation methods.
Engineers considering using USB to integrate computer based measurement systems need to
understand isolation issues and how to deal with them before choosing their measurement
hardware. They need to understand isolation issues so they can understand how external ground
connections interact with, not only the computer hardware, but analog signal integrity as well.
Isolation protects your PC from damage and preserves the integrity of the data by physically
separating electrical connections between circuits, thereby eliminating potentially harmful voltage
or current from flowing through your system.
As Figure 1 shows, a data acquisition module, whether based on USB or not, consists of two
sections: a measurement section containing the analog front end and digitizing electronics, and a
data communication section that transfers those signals to the host computer. In a non-isolated
system, the measurement and data communications sections are both referenced to the same
ground potential the computers chassis ground. Designers of isolated measurement systems
go to great lengths to ensure that there is no direct current path between the measurement and
data-communications sections.
Figure 1: Data acquisition modules, whether built for PCI, USB or any other bus standard, have a
communications section and a measurement section.
The USB standard does not mandate isolation. To do so would drive up costs unnecessarily.
There is nothing in the standard requiring, promoting, or even suggesting that peripherals
connected to the computer via USB be isolated.
The engineers who developed and now promote USB, however, know how important isolation
can be for many potential applications including computer-based instrumentation. In all of their
seminars, technical writings and other communications, they strongly recommend isolation for
non-Human Machine Interface (HMI) peripherals.
Potential Gotchas
The USB signals are on the order of 800 millivolts differential, which makes them low voltage
differential signals (LVDS). As such, they are subject to interference from various external
sources. These sources dont cause problems for highly-shielded HMI applications, but they can
trip up measurement systems that necessarily must electrically touch Units Under Test (UUTs)
in the uncontrolled outside world. A mouse or key board is isolated by the plastic case.
There are five general classes of interfering phenomena that isolation helps shield your
measurements from:
Electrostatic Discharge (ESD) is any discharge from a static source into your measuring
equipment. Typically what happens is the application locks up and you have to close it down and
restart it. If you have a gross failure, you can blue-screen your system or worse damage your
hardware. International product standards, such as CE ratings, mandate ESD tolerance up to
4,000 Volts for products with exposed metal and air discharge of 8,000 volts for shrouded or
plastic devices.
Ground Ambiguity results from signals in different circuits being referenced to different grounds.
Even a data acquisition card has separate analog grounds, digital grounds, chassis ground (for
the shielded cables), and power ground. When connecting your measurement system to the
outside world, there is no guarantee that the UUT wasnt mis-wired enough to apply 110 or 220
Vac into your analog ground or I/O signals. Even knowing theres a solid wire between two
ground points is no guarantee. Current flowing through a power-ground conductor can raise the
voltage at the hot end enough to compromise your measurements or destroy your computer.
Ground Loops appear when there are multiple paths for signal and power supply currents to
return to the power supply ground. Long cables act like antennas magnetically picking up any
low-frequency electric fields that happen to be wandering around the neighborhood, as Figure 2
shows. Fields generated by electric motors, transformers, and even building wiring carrying
power to run the microwave oven in the break room generate significant fields that ground loops
can couple into your measurement system.
Figure 2: Ground loops can appear when spatially separated pieces of equipment share a
common ground.
Ground and Common-Mode Noise arise when any of a number of effects push random currents
into your signal reference, lifting both the signal high and signal low in unison, as Figure 3 shows.
In extreme cases, surges large enough to damage equipment can occur. A perfect example of
low-level common-mode noise appears when your analog signal shares power or ground with a
large number of active logic gates or even worse relays with a potential back EMF. Each time a
circuit opens or closes; it changes the load on the power supply and the current returning through
the ground system. These rapid current variations induce voltage spikes that can amount to as
much as a hundred millivolts across a circuit-board trace enough to disrupt sensitive
measurements.
EMI/RFI are extraneous high-frequency (tens of kilohertz on up) signals picked up by your analog
measurement front end from long cables acting like antennas picking up any electric fields that
happen to be in the neighborhood. While low frequency noise can only gain entry by magnetic
induction, high-frequency fields can reach in through capacitive, inductive or conductive coupling.
Once inside, they can jump from circuit element to circuit element. Not only can they play havoc
with your measurements signal-to-noise ratio, they can get into your processor. If that on-board
processor hangs, your measurement system can lock up. Youll lose communication with the
USB, and thats when your application hangs!
Figure 3: Common-mode voltages move both sides of a balanced signal transmission line relative to ground.
Isolation Methods
Isolation refers to setting up a barrier to electrical current between one part of a device and
another, while allowing certain interactions (e.g., signals and power) to pass. An extreme
example is the plastic case and keys on your keyboard. They place an insulating barrier (the
plastic) between your hands and the electric currents and voltages that make the keyboard work,
while allowing mechanical signals (key presses) to pass.
To stop interference from the five phenomena listed above, isolation is needed between the
measurement and data communication sections shown in Figure 1. The isolation barrier must
allow power, control signals and data to pass, while decoupling the ground systems.
Figure 4 shows three primary methods of isolating measurement systems. Each has its particular
advantages and disadvantages.
Figure 4: Three widely used isolation methods use non-electrical means to couple signals across
an insulating gap: optoisolators use light; transformers use magnetic fields; and differential
capacitive couplers use electric fields.
Optoisolators combine a light-emitting diode with a photosensitive transistor to send signals
across an isolation gap in the form of light. These devices are especially useful for passing digital
signals, but not power. They have a particular advantage in that they do not use any power when
in their off state. This is important considering that isolated power is expensive and supply
efficiency is typically 75%. Optoisolators, however, introduce propagation delays on the order of
tens of microseconds as well as significant jitter.
Transformers provide isolation via magnetic coupling. Theres no DC electrical path through a
transformer. Transformers can pass enormous amounts of power, especially at high frequencies,
and do so fairly efficiently, so they are particularly useful for providing isolated power or any
highly repetitive signal, such as clock signals. Transformers also introduce no jitter. They are,
however, relatively large and heavy compared to integrated circuit components and are not
amenable to solid-state fabrication techniques. Additionally, the transformer is a two way street
for noise pick-up.
Differential capacitor coupling (DCC) makes use of the fact that capacitors are quite good at
passing AC signals, but simply will not pass direct current. The major drawbacks to DCC is
propagation delay and jitter. DCC is excellent for isolating high-speed digital data paths.
Isolation Improves Measurements
Figure 5 shows a representative architecture for an isolated USB module. The data
communications side has a USB chip along with a processor that mediates the communications
from the host computer across the isolation barrier to the measurement section. A second
processor (a CMOS programmable logic controller CPLD) controls the data acquisition activity
in the measurement section.
Figure 5: To isolate a USB measurement module, designers put an isolation barrier between the data
communication and measurement sections, and add minimal processing power on the isolated side.
Before anything can happen on the measurement side of the isolation barrier, you have to get
electrical power across to energize the circuitry. A switching power supply (not shown in the
figure) uses transformer coupling to accomplish this task. On the non-isolated side, a DC-to-AC
converter (basically an oscillator running at a few hundred kilohertz) provides AC power to pass
through the transformer. On the isolated side, a rectifier and filter convert the AC power back to
DC.
With the measurement circuitry energized, the non-isolated processor can set up the
measurement parameters, such as gain, sample rate, channel scanning, etc. and load them into
the CPLD memory. It sends these control and set-up signals via optoisolators.
Finally, the measurement system acquires the data. The analog section makes the
measurements, which the digitizer converts to digital signals for the CPLD to pass back over the
isolation barrier via DCC.
On the non-isolated side of the barrier, the processor takes that data and passes it on to the host
computer via USB. The data arrives safely and in good condition because the isolation barrier
effectively decouples the measurement ground from the computers power supply and chassis
ground.
With the computer and measurement grounds isolated, you are free to reference your
measurements to the proper point on the UUT, or even allow it to float. Should the UUT receive
an ESD jolt, for example, the isolation system allows the measurement ground to float up with the
UUT ground until the UUTs static-dissipation system bleeds off the charge. If the UUT doesnt
have a static-dissipation system, the isolation barrier prevents that failing from affecting the
measurements or damaging anything in the measurement system.
Ground ambiguity is eliminated because the measurement reference can now float with the UUT
reference. Similarly, ground loops are less of a problem because USB allows you to move the
analog front end closer to the UUT, eliminating the need for long signal leads.
While isolation wont eliminate common mode voltages, it allows the measurement reference to
move with them. The voltages are still there, however, they dont affect the measurements.
It Gets Better!
Isolating your measurement system lets you apply measurement techniques that are not possible
with non-isolated measurement systems. Figure 6, for example, shows a situation where you
want to measure the voltage output of, say, the fortieth in a stack of eighty 1.25 VDC fuel cells.
Figure 6: Isolation allows you to make floating-ground measurements instead of differential
measurements referenced to ground.
Non-isolated measuring equipment would force you to make a differential measurement between
48.75 V and 50 V. The digitizers dynamic range would have to be better than 50 V. Assuming
that the analog front end has a calibrated gain setting for 50 V and the digitizer has 12-bit
resolution, the digitization error is 0.012 V, or 1% of the measurement. (50/4096 = 0.012V)
Having isolation in your USB module allows you to use a single-ended floating-ground technique
with the analog ground connected below the 40th cell. The dynamic range only needs to be at
least 1.25 V. The analog front end above would likely have a 5 V gain setting available. Using it
would provide a digitization error of 0.0012 V, or 0.1% error.
Similarly, isolating your measurement section allows you to apply a host of classic in-circuit
measurement techniques, such as guarding, 4-wire Kelvin, back driving and so forth, which just
arent possible otherwise.
Engineers wanting to use USB to integrate their computer-based test and measurement systems
cannot take measurement-circuit isolation for granted. It is up to you to ensure that the USB data
acquisition module you select incorporates isolation technology. Because isolation is expensive to
engineer into a module, companies that make the investment are motivated to tell you about it.
So, look at the specifications. If the literature doesnt say its isolated, it probably isnt. Dont get
zapped!
Why consider USB for computer based measurements?
Expansion slots in personal computers are rapidly going away, while USB ports are multiplying.
Just a few years ago, a new PC would come equipped with, if you were lucky, two USB ports.
Today, they typically arrive with four or more. This reason alone is enough for users of test and
measurement equipment to consider switching from PCI to USB to interconnect their computerbased
instrumentation system components.
In addition, there are a number of USB features that make it desirable as an instrumentation
integration standard:
Plug-and-play hardware connection You dont have to open your PC any more to plug an
expansion card. You simply plug your USB module into the external port. You dont have to
power it down!
Keep measurement circuitry out of the noisy environment inside the PC case This is a very
important consideration when you consider that you now have processors in the PC case going at
over a gigahertz that look like pure noise sources to an analog data acquisition front end.
Move the measurement circuitry closer to the UUT Putting the instrumentation analog front
end on a long (up to 5 meters) cable allows you to shorten your interconnect wiring to the UUT.
High-speed data transfer Maximum data transfer rate is 480 Mbps for Hi-Speed USB 2.0.
Consumer-oriented standard helps drive down cost Taking advantage of the fact that that
USB chips are manufactured for the consumer market drastically reduces component costs for
instrumentation equipment manufacturers. They are then able to pass these savings on to their
customers in the form of lower prices.
Easy portability USB allows test engineers to move instrumentation equipment from location
to location more easily. Instead of having to move entire host computers, they just unplug the
USB module and plug it into a computer at the new location.
Easy expandability Most currently available desktop PCs have more USB ports available than
PCI or ISA expansion slots. In addition, inexpensive external USB hubs are available from
consumer electronics stores, allowing you to expand your USB-based instrumentation system to
up to 127 devices.
Simple power connections The USB specification mandates that the host provide 5 VDC power
at up to a total of 500 mA to peripherals. Many external hubs add to the systems current capacity
as well.
Laptops and Isolation
If you use a laptop computer as your data acquisition host, and dont plug it in to change the
batteries while making your measurements, you have an isolated system, even if your USB
measurement module is non-isolated. Whatever ground you connect your module to becomes the
reference potential for the entire laptop. Keep in mind, though, if you plug any peripheral, such as
a printer, into the laptop you ruin that isolation.