Cole-Parmer Technical Library
Gravity Feed Installation
of your Centrifugal Pump
This illustration shows a typical setup for a gravity feed system. Since most centrifugal pumps require priming, be sure to position pump at the bottom of the supply tank and retain a liquid level higher than the pump inlet port.
You can regulate the output flow with a ball valve or a flowmeter with a control valve on the discharge line.
Related Links: Printable page E-mail a colleague
Complete Selection of Pumps - Centrifugal Pumps
Centrifugal Pump Selection Guide
General Pump FAQs
Guide to Liquid Pumps
Liquid Pumps General Information
Pump Types and Definitions
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data