Cole-Parmer Technical Library
Horsepower Requirements for Pumps
Use this chart as a general guideline for sizing a motor for pedestal-mount or pulley-driven pumps. Other factors (such as viscosity, particulate matter, etc.) may require increased horsepower. Use individual pump curves (when given) for more precise calculations.
GPM Horsepower required to lift water (at 60°F) to a given height
5 ft 10 ft 15 ft 20 ft 25 ft 30 ft 35 ft 40 ft 50 ft
5 0.006 0.013 0.019 0.025 0.032 0.038 0.044 0.051 0.063
10 0.013 0.025 0.038 0.051 0.063 0.076 0.088 0.10 0.13
15 0.019 0.038 0.057 0.076 0.095 0.11 0.13 0.15 0.19
20 0.025 0.051 0.076 0.10 0.13 0.15 0.18 0.20 0.25
25 0.032 0.063 0.095 0.13 0.16 0.19 0.22 0.25 0.32
30 0.038 0.076 0.11 0.15 0.19 0.23 0.27 0.30 0.38
35 0.044 0.088 0.13 0.18 0.22 0.27 0.31 0.35 0.44
40 0.051 0.10 0.15 0.20 0.25 0.30 0.35 0.40 0.51
45 0.057 0.11 0.17 0.23 0.28 0.34 0.40 0.46 0.57
50 0.063 0.13 0.19 0.25 0.32 0.38 0.44 0.50 0.63
60 0.076 0.15 0.23 0.30 0.38 0.46 0.53 0.61 0.76
70 0.088 0.18 0.27 0.35 0.44 0.53 0.62 0.71 0.88
80 0.10 0.20 0.30 0.40 0.50 0.61 0.71 0.81 1.0
90 0.11 0.23 0.34 0.46 0.57 0.68 0.80 0.91 1.1
100 0.13 0.25 0.38 0.50 0.63 0.76 0.88 1.00 1.3
Related Links: Printable page E-mail a colleague
Complete Selection of Pumps - Centrifugal Pumps
Complete Selection of Pumps - Drum and Hand Pumps
Complete Selection of Pumps - Gear Pumps
Complete Selection of Pumps - Ismatec
Complete Selection of Pumps - Manostat Peristaltic Pumps
Complete Selection of Pumps - Masterflex Peristaltic Pumps
Complete Selection of Pumps - Metering Pumps
Complete Selection of Pumps - Postive Displacement Pumps
General Pump FAQs
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Terminology
Liquid Pumps Technical Equations and Information Tables
Pump Types and Definitions
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data