|
For tubing that lasts a long time, provides purity, and features excellent temperature and chemical resistance,
an ideal choice is Norprene® Food pump tubing from Masterflex®. Here's why:
 |
Ultra long tubing life (over 10,000 hours when used in a Masterflex® tubing pump!) |
 |
Meets FDA, 3-A, and NSF criteria |
 |
Ideal for pumping viscous fluids |
 |
Non-aging, nonoxidizing and ozone resistant |
 |
Heat sealable and bondable |
 |
Temperature resistant from -60ºF to 275ºF |
 |
Excellent chemical resistance |
 |
Repeatedly autoclavable |
Norprene Food tubing consistently outperforms general purpose tubing in a variety of applications. It will not weaken or crack
following years of exposure to heat and ozone. It is temperature resistant up to 275ºF and retains flexibility down to -60ºF. It provides
superior resistance to many corrosive fluids, and offers excellent acid and alkali resistance.
This tubing fully complies with FDA 177.2600 criteria, 3-A Sanitary Standards, and the NSF Standard 51, which are applicable in many food contact
applications. And to ensure you receive a superior quality product that will perform to specifications, all Masterflex®
tubing is optically inspected and factory tested.
|
|
Typical Applications
|
| Cosmetic production
Food and beverage
Water purification
|
Printing
Chemical transfer
Semiconductor manufacturing
|
Masterflex Norprene Food Tubing
Masterflex Norprene Food Spooled Tubing 06415-13
|