Cole-Parmer Technical Library
Masterflex® Connection Newsletter
Volume 4, Number 2, 2002
QUESTIONS & ANSWERS . . .
I/P® DIGITAL PROCESS DRIVE 77420-00
Q: What applications is the new I/P® Digital Process drive designed for?
A: It's almost endless, from industrial to lab process, pharmaceutical and chemical processing to wastewater. The fact is, when your application requires a reliable, easy-to-use drive that produces high flow rates, this one's ideal.
Q: It sounds very versatile.
A: Yes, and it's compatible with all I/P pump heads and every Masterflex tubing formulation, so you can build just the pump you want. You can even stack two Standard or Easy-Load pump heads.
Q: Why stack pump heads?
A: By adding a second pump head to the drive, you'll increase the maximum flow rate to 26 LPM.
Q: Is it difficult to use?
A: Not at all, especially if you have used Masterflex pumps before. The I/P digital drives operate similarly to our L/S pumps. If you're scaling up, you'll already know how to use this drive. Even if you've never used a Masterflex pump before, this one's designed with ease of use in mind.
Q: How durable is it?
A: Very. Constructed of seamless 316 stainless steel, the I/P digital drive features precision bearings and a heavy-duty keypad interface. With no gear box, it's quiet, efficient, and can be run seven days a week, 24 hours a day.
Q: Is it low maintenance?
A: Yes, it features direct drive technology and a brushless, reversible motor that's maintenance-free.
Q: How about clean-ups?
A: This IP55-rated wash-down drive with a 316 stainless steel housing is perfect for harsh environments. After use, simply hose the unit down to clean.
Masterflex I/P® drive
77420-00
shown with an I/P
High-Performance
pump head
Related Links: Printable page E-mail a colleague
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data