Cole-Parmer Technical Library
Maximum Holding Times
for Perishable Foods
Temp (°F) Safety* Quality**
100*** 4 hr 2 hr
95*** 5 hr 2.5 hr
90*** 6 hr 3 hr
85*** 7 hr 3.5 hr
80*** 8.5 hr 4 hr
75*** 11 hr 5.5 hr
70**** 14 hr 7 hr
65**** 18 hr 9 hr
60**** 1 day 12 hr
55**** 1.5 days 18 hr
50**** 2.5 days 30 hr
45**** 5 days 2.5 days
41 10 days 5 days
40 13 days 6.5 days
35 30 days 15 days
32 60 days 30 days
30 Safe chilled food holding
28 Meat, poultry, fish thaw
23 Spoilage bacterial begin to multiply
14 Yeasts and molds begin to multiply
*10 multiplications of pathogens.
**5 multiplications of pathogens
***Applied heat for less than 6 hours.
****Cooled for less than 15 hours
Note: References for technical data available upon request.
Related Links: Printable page E-mail a colleague
Complete Selection of Dataloggers
Complete Selection of Food Quality
Complete Selection of Food Safety
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data