Cole-Parmer Technical Library
Masterflex® Pharmaceutical Applications
Masterflex® pumps are designed to be non-contaminating and non-invasive, making them the pumps of choice in the pharmaceutical industry. Easy-Load® pump heads and a wide range of drives fit many applications, from pumping nutrients or pH adjusters for fermentation, to dispensing cosmetics into small containers. The tubing is traceable to USP Class VI and can be easily sterilized.
Applications-Partial list
Agar dispensing
Aspiration of tissue culture medium
Bioprocess container transfer
Circulation of cell suspension in fermentation
Cosmetic dispensing
Dispensing agar into petri dishes
Dissolution testers
Fermentation control
Harvesting cell media
Manufacturing IV bag dispensing
Nutrient supply for cultures
Spray coating dispensing
Sterilized media dispensing
Ultrafiltration
FEATURES BENEFITS
Wide range of tubing materials Compatible with many laboratory chemicals.
Non-contaminatingFluid media only contacts ID of tubing.
Easy clean-up Just change the tubing to clean the pump
Multichannel capabilityUse up to 8 channels simultaneously on one drive.
Sanitary connections on tubing Quick tubing connections within sanitary systems.
Handles viscous materials Dispense agars and suspensions.
Runs dryUse as a low flow aspirator or air/vacuum pump.
Modular design Mix and match pump heads, tubing and drives.
OEM packagesDesign into your lab dispensers or analytical equipment.
Related Links: Printable page E-mail a colleague
Complete Selection of Pumps - Masterflex Peristaltic Pumps
http://www.coleparmer.com/catalog/bioconnect/bioconnect_index.asp
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data