How to Read a Hydrometer
- Pour your sample into a smooth, clear cylinder or jar that is dry or well rinsed with a portion of the sample.
- Make sure your sample is thoroughly mixed before testing.
- Immerse the hydrometer in the liquid to a point slightly below the place where it naturally floats. Make sure the hydrometer and liquid are at rest and free of air bubbles.
- Measure the temperature of the sample. Ideally, the sample temperature should be equal to the temperature standard of the hydrometer, generally 60°F. If temperature differences are unavoidable, correction tables can help to adjust readingscall our Applications Specialists for information.
- Take your reading at the point where the surface of the liquid crosses the hydrometer.
Did You Know...
| Degrees Baumé = 145- | 145 sp gr 60°F / 60°F |
| Degrees API = | 141.5 sp gr 60°F / 60°F | -131.5 |
Complete Selection of Hydrometers
Hydrometer Selection Guide