Cole-Parmer Technical Library
Chiller/Recirculator Selection Guide
Use the following formulas to determine the cooling capacity (in watts) required by your system. We recommend selecting a chiller/ recirculator that has a reserve capacity of 10-20% to compensate for radiant losses through tubing and to the environment.
Immersion Chillers
Watts =
V • ΔT • 70 • k
t
V = total liquid volume (liters)
ΔT = temperature difference (°C)
t = heating/cooling time (minutes)
k = specific heat constant
k = 1.0 for water
k = 0.4 for silicone bath fluid 01296-00
k = 0.5 for alcohols
Flow-Through Chillers and Recirculators
Btu
hr
= flow rate (gph) • Gc • k • ΔT
(note:
Btu
hr
= watts x 3.41)
Gc = weight of coolant (lb/gallon)
   Water: 8.34 lb/gallon
   Silicone bath fluid 01296-00: 7.09 lb/gallon
   Ethylene glycol/water 01294-30: 7.5 lb/gallon
k = specific heat constant (Btu/lb °C)
k = 1.0 for water
k = 0.4 for silicone bath fluid 01296-00
k = 0.5 for ethylene glycol/water 01294-30
ΔT = temperature difference (°C)
Related Links: Printable page E-mail a colleague
Complete Selection of Baths and Circulators
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data