  Cart  Subtotal = 0.00
1-888-358-4717 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Cole-Parmer Technical Library
Safety Gloves Selection Guide
Glove Features
Supported or Unsupported Gloves
Unsupported Gloves are made from hand forms that are dipped directly into a glove compound with no supporting liner—provide good tactility and dexterity.
Supported Gloves have a knitted or woven cloth liner that adds structural strength. These gloves offer greater resistance to snags, abrasions, cuts, and punctures but do not have the tactility of unsupported gloves.
Grip Design
Grip refers to embossed patterns or finishes on the palm and fingers that allow hand traction on objects being handled in wet or dry conditions. Patterns are embossed on the surface of the glove material, or the material can be finished with various rough textural configurations such as pebbling or crinkling.
Cuff Design
Pinked Cuff has a zigzag appearance; catches drips in cuff.
Rolled Cuff acts as a barrier to keep chemicals from running off glove onto skin.
Straight Cuff protects skin from chemical run-off.
Knitwrist Cuff absorbs perspiration and adds temperature protection.
Gauntlet Cuff has flared design and extended length to protect wrist and forearm.
Related Links: Printable page E-mail a colleague
Complete Selection of Gloves
Safety Glove Chemical Compatibility Database
Safety Gloves Sizing Guide
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data