Cole-Parmer Technical Library
Vacuum/Compressor Pump Selection Guide
Pump Types
Dry Pumps (nonlubricated) provide a clean vacuum over the entire vacuum range. Dry pumps require little maintenance and don't generate any waste material
Water-Lubricated Pumps provide a rough vacuum at low initial cost and are very simple to operate. These pumps are corrosion resistant but are not suitable for water vapor sensitive systems
Oil-Lubricated Pumps offer high capacities, high vacuum/pressure levels, and lower noise levels. Diffusion pumps feature an ultra-high vacuum. Oil-lubricated pumps, however, require routine maintenance and oil changes, and the oil may contaminate your vacuum
Vacuum/Pressure Pumps Selection
To determine which pump is right for your application, calculate your free-air capacity by adding up the free-air requirements for all air-powered instruments. Add an additional 25% to allow for system variables and to provide capacity for future needs. Then determine the maximum vacuum or pressure required for your application—see the "Vacuum/ Pressure Conversion Table" below for any necessary values.
Vacuum/Pressure Conversion Table
Absolute vacuum Relative vacuum
torr/mm Hg mbar psia atm % vacuum " Hg
760 1013 14.696 1.0 0.0 0.0
750 1000 14.5 0.987 1.3 0.42
500 667 9.7 0.658 34 10.22
400 533 7.7 0.526 47 14.22
300 400 5.8 0.395 61 18.12
200 267 3.9 0.264 74 22.07
100 133.3 1.93 0.132 87 25.98
70 93.4 1.35 0.0921 90.8 27.16
50 66.7 0.97 0.0658 93.5 27.95
25.4 33.8 0.4912 0.034 96.6 28.92
10 13.33 0.193 0.0132 98.7 29.53
1 1.33 1.934 x 10-2 1.32 x 10-3 99.9 29.88
0.1 0.133 1.93 x 10-3 1.32 x 10-4 99.99 29.916
1 x 10-2 1.33 x 10-2 1.93 x 10-4 1.32 x 10-5 99.999 29.9196
1 x 10-3 1.33 x 10-3 1.93 x 10-5 1.32 x 10-6 99.9999 29.91996
1 x 10-4 1.33 x 10-4 1.93 x 10-6 1.32 x 10-7 29.91999
1 bar = 14.5 psi
1 cubic foot = 28.32 liters
1 micron (1µ) = 0.001 mm
(29.92 - " Hg) x 25.4 = torr/mm Hg
1" H2O = 0.0361 psi
1" H2O = 0.0735" Hg
Did You Know:
For every 1000 feet above sea level, the listed maximum vacuum for your pump will be reduced by 1" Hg.
Related Links: Printable page E-mail a colleague
Complete Selection of Vacuum Pumps
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data