| Phase Desription |
Similar Phase |
Similar column |
| Non-polar |
|
|
| Dimethylpolysiloxane |
OV-1, OV-101, SE-30 |
| DB-1 |
SPB-1 |
HP-1 |
RTx-1 |
CPSil 5CB |
AT-1 |
BP-1 |
|
(5% diphenyl) dimethylpolysiloxane |
SE-52, SE-54, OV-73 |
| DB-5 |
SPB-5 |
HP-5 |
RTx-5 |
CPSil 8CB |
AT-5 |
BP-5 |
|
| Midpolarity |
|
|
(14% cyanopropylphenyl) methylpolysiloxane |
OV-1701 |
| DB-1701 |
SPB-1701 |
|
RTx-1701 |
CPSil 19CB |
AT-1701 |
BP-10 |
|
(50% phenyl) methylpolysiloxane |
OV-17 |
| DB-17 |
SPB-2250 |
HP-50 |
RTx-50 |
|
AT-1701 |
|
|
| Polar |
|
|
(50% cyanopropylphenyl) methylpolysiloxane |
OV-225 |
| DB-225 |
SPB-2330 |
HP-225 |
RTx-225 |
CPSil 43CB |
AT-225 |
BP-225 |
|
| Polyethylene glycol(PEG) |
Carbowax 20 M |
| DB-WAX |
SUPEL COWAX 10 |
HP-(iNNO)Wax |
Stabil wax |
CP-Wax 52CB |
AT-WAX |
BP-20 |
|
(78% cyanopropyl) methylpolysiloxane |
Silar 7CP,SP-2310 |
| DB-23 |
SP-2230 |
|
RTx-2330 |
CPSil 88CB |
AT-Silar |
BPX-70 |
|