Cole-Parmer Technical Library
Temperature Conversion Equations
°F = (1.8 x °C) +32
°C = (°F-32) x 0.555
Kelvin = °C + 273.2
°Rankin = °F + 459.67
Related Links: Printable page E-mail a colleague
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data