| Cole-Parmer Technical Library | |
|
|
|
The diagram at right shows the accuracy versus temperature for a typical thermocouple meter and probe. For example, using the diagram, the meter error at 250°C is ±0.5°C and the probe error at 250°C is ±1.0°C, thus the system accuracy would be ±1.5°C or better. The result is often conservativethe actual system accuracy is usually better. Glass thermometers, labels, paints, bimetal thermometers, hand-held infrared thermometers, and digital indicators include a general system accuracy statement. National Institute of Standards and Technology (NIST) documentation gives you the actual system accuracy. Knowing the actual system accuracy gives you the ability to compensate or calibrate your instruments measurements for greater accuracy. View our complete listing of our NIST temperature calibration services
Complete Selection of Thermocouple Instruments Complete Selection of Thermocouple Probes Temperature Conversion Equations Temperature FAQs Temperature Instrument Ranges -Thermocouple Selection Guide |
Product Selection Guides | Product Maintenance | FAQs | Technical Library Map
Articles and White Papers | Case Studies | Conversion Factors and Tech Data