Viscosity Values
Centipoise* (cp) | Centistokes (cSt) | Saybolt Second Universal (SSU) | Typical liquid |
| 1 | 1 | 31 | Water |
| 3.2 | 4 | 40 | Milk |
| 12.6 | 15.7 | 80 | No. 4 fuel oil |
| 16.5 | 20.6 | 100 | Cream |
| 34.6 | 43.2 | 200 | Vegetable oil |
| 88 | 110 | 500 | SAE 10 oil |
| 176 | 220 | 1000 | Tomato juice |
| 352 | 440 | 2000 | SAE 30 oil |
| 880 | 1100 | 5000 | Glycerine |
| 1561 | 1735 | 8000 | SAE 50 oil |
| 1760 | 2200 | 10,000 | Honey |
| 3000 | 4500 | 20,000 | Glue |
| 5000 | 6250 | 28,000 | Mayonnaise |
| 8640 | 10,800 | 50,000 | Molasses B |
| 15,200 | 19,000 | 86,000 | Sour cream |
| 17,640 | 19,600 | 90,000 | SAE 70 oil |
Complete Selection of Viscometers
Common Viscometer Terms