Cole-Parmer Technical Library
Sterilized Disposable Bioprocess Consumables
The latest trend sweeping the Biopharmaceutical market!
Sterile Disposable Bioprocess Filling System
Click image to see animation
click to see animation
One of the most basic applications for sterilized disposable bioprocess assemblies is mixing/filling/storage. In the above illustration, biologics are blended, then pulled from the mixer through sterile tubing using a peristaltic pump. After passing through a .2u sterilizing filter, the material is collected in sterile bio-bags, which are eventually separated off from the system by heat sealing the filling lines. The remainder of the assembly is discarded after use.
They are the biopharmaceutical industry equivalent of the iPod®—the hottest thing to hit the market since the peristaltic pump. They are sterilized disposable bioprocess system assemblies, and if your company hasn’t yet seriously considered them as an alternative to stodgy, old 20th century technology, rest assured your competition has.
Why have disposable bioprocess consumables become so popular? Because they save time and production cost while typically improving quality.
Previously, when a company went into biologic production they had to purchase components like filters, tubing, valves, and storage containers from multiple vendors. Then cleanroom support teams would have to spend time assembling, sterilizing, and validating the components before even beginning production.
Purchasing custom-ordered, sterilized, and preassembled bio-production assemblies such as those now available through Cole-Parmer’s new BioConnect™ program, allows companies to limit downtime in their plants, reduce the amount of facility space needed for bioprocess, and eliminate a substantial portion of their cleaning and validation requirements. In other words it allows them to avoid what is often the most costly and complex, validation-intensive portion of the process.
What applications can these integrated disposable solutions be applied to? Media filtration, cell culture supplements, clarification, separation, ultrafiltration, and final fill just to name a few. They are ideal for pharmaceutical and biologic drug production, and for the storage and shipping of any sterile-filtered fluids. They also have many potential uses in the food & beverage market.
At their most basic, these are simple, closed systems which create a Class Zero environment—meaning cleanrooms are not required in order to maintain sterile operation. No environmental or clean air controls are necessary.
The assemblies are available either sterile or non-sterile. Sterilization is most often accomplished through gamma irradiation, as is the case with BioConnect assemblies. All custom-designed and custom-assembled units produced by BioConnect are irradiated with a minimum of 25kGy to assure a sterility assurance level (SAL) of 10-6, which represents a critically acceptable one-per-million chance of microbial contamination. Sterilized units are then shipped to the end user, where they are installed and can be put immediately into operation.
Cole-Parmer, through the BioConnect program, is the first company to validate the sterility of the entire assembly. While competitors validate the sterility of bags and filters separately before assembly, with BioConnect the entire system—from tubing to filters to individual connections—is assembled then validated for sterility to the 10-6 SAL. Our selection of fluid handling products, filling systems, filtration equipment, and process containers is second to none. All assemblies have full lot traceability and are manufactured in a Class 10,000 cleanroom. Our facility is ISO-13485-certified, cGMP-compliant and FDA-registered.
You may need to download Flash to see the above animation.
Related Links: Printable page E-mail a colleague
Complete Selection of Tissue and Cell Culture
http://www.coleparmer.com/catalog/bioconnect/bioconnect_index.asp
Cole-Parmer’s Bioconnect Program
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data