Characteristics of Sanitizers
| Characteristics | Steam | Chlorine | Iodophor | QUATS* | AAS** |
| Gram-positive bacteria | Best | Good | Good | Good | Good |
| Gram-negative bacteria | Best | Good | Good | Poor | Fair |
| Spores | Good | Good | Poor | Fair | Fair |
| Yeasts and molds | Best | Good | Good | Fair | Poor |
| Bacteriophage | Best | Good | Fair | Poor | Poor |
| Optimum pH | N/A | 4-5 | 3 | 10 | 2-2.5 |
| Penetration | Poor | Good | Poor | Good | Excellent |
| Corrosive | No | Yes | Slight | No | Slight |
| Irritation | N/A | Yes | No | No | Yes |
| Hard water affects | No | No | Slight | Yes/No | Slight |
| Shelf-life | N/A | Short | Long | Long | Long |
| Organic matter affects | No | Yes | Slight | Low | Slight |
| Stable in 140°F H2O | N/A | No | No | Yes | Yes |
| Leaves residue | No | No | Yes | Yes | Yes |
| Flavor/odor | No | Yes | Yes | No | No |
| Ease of use | Poor | Excellent | Excellent | Foam | Foam |
| Use with detergent | No | No/yes | Yes | No/yes | No |
| Maximum use by FDA/USDA | None | 200 ppm*** | 25 ppm**** | 200 ppm***** | 200-400 ppm |
| Cost | High | Low | Moderate | Moderate | Moderate |
| Where to use | Equipment | Equipment, floors, walls | Rubber, plastic crates | Aluminum, walls, floors | Stainless steel, CIP |
*Quaternary ammonium compounds. **Acid anionic surfactants.
***Nonfood contact use on concrete floors (1000-5000 ppm), conveyor belts (300-500 ppm), fogging with 800-1000 ppm, wooden crates (1000 ppm).
****Nonfood contact use on concrete floors (500-800 ppm).
*****Nonfood contact use on cooler walls (500-800 ppm), concrete floors (500-800 ppm).
ChemicalsFoodborne Pathogens
ChemicalsHow I Survived a Product Recall!
ChemicalsNew Templatre
ChemicalsTypes of Cleaning Compounds