Cole-Parmer Technical Library
Cole-Parmer eNEWS April 2009
To view this e-mail as a Web page, copy and paste the following link into your browser’s URL Address window: http://www.coleparmer.com/techinfo/techinfo.asp?referred_id=4357&htmlfile=e-news_LAB_04-09.htm
eNEWS you can use
April 2009        
Request your FREE 2009/10 General Catalog today!
Ultrasonic Fractionation
of Cells in Microplates
multi-element probes
Ultrasonic processors are now widely used in laboratories for a variety of applications. The use of 4-element probes increased productivity and improved repeatability over single probes. The recent addition of 8- and 24-element probes meets the high-throughput requirement of genomics and proteomics laboratories. Read how these time-saving multi-element probes will favorably impact your workload while ensuring uniform and predictable results.
Full Article
Cole-Parmer Ultrasonic Processors
Multi-Element Probes
An Invaluable Tool for Water Analysis
 electrode handbook
This comprehensive resource covers the full range of Thermo Scientific pH electrode families including the best-selling ROSS Ultra®, ROSS®, and PerpHect® electrodes and the environmentally friendly “green” electrodes. All the information you need to operate, maintain, and troubleshoot your pH electrodes.
Download Free Handbook
(1MB pdf)
Thermo Scientific Orion Laboratory pH Electrodes
Tech Challenge
Take the challenge,
see if you’re right...
Chemical Grades—Not All the Same
Challenge: A customer calls in to inquire why their HPLC column is no longer lasting as long as it normally does, and informs the Application Specialist that the cost of replacing the column will throw off their budget if the issue continues. Their work load and test procedure have not changed, so they are wondering why there has been such a difference.
The customer uses the HPLC to test for the caffeine level in sodas they manufacture and there is no specific FDA test procedure they need to follow during testing. However, their results do have to be within a certain range, even though the exact caffeine level is not listed on the product label either. After some investigation, it was discovered that in trying to save money the customer had changed to a cheaper chemical. They had gone from using an ACS grade to a chemically pure grade; the customer assumed since it stated it was pure it would work similarly. What is the lowest chemical grade they could use in their application to save money and perform efficiently in their instrument?
  1. Certified ACS—A chemical grade of highest purity; it meets or exceeds purity standards set by the American Chemical Society (ACS). Other grades at this level (dependent upon the manufacturer) include HPLC grade, ACS Plus, GC Resolv®, Spectranalyzed®, and Optima™.
  2. Reagent/ACS grade—High purity generally equal to certified ACS grade and suitable for use in many laboratory and analytical applications. However, it has not established certification with ACS requirements.
  3. USP—A chemical grade of sufficient purity to meet or exceed requirements of the U.S. Pharmacopeia (USP); acceptable for food, drug, or medicinal use; may be used for most laboratory purposes. This grade may also be listed as NF/FCC/EP/BP/JP certified dependent upon the manufacturer and popular markets for the chemical.
  4. Lab grade—For use in routine laboratory applications as well as histological purposes
See the answer.
Our technical experts answer questions like this every day. Get the answers to your questions by calling our Application Specialist at 800-323-4340.
Echotherm digital hot plates
What You Should Know about Temperature Control and Hot Pates
In general, the better the temperature controller is in your hot plate, the smaller the temperature cycle and the temperature overshoot or undershoot values. Learn why all EchoTherm® digital hot plates and stirring hot plates feature PID temperature control.
Full Story
EchoTherm Digital Hot Plates
EchoTherm Digital Stirring Hot Plates
EchoTherm Programmable Digital Stirring Hot Plates
New Masterflex L/S Digital Drives
Featured Product!
New Masterflex® L/S® Digital Drives
Precision pump drive offers a maintenance-free high-accuracy motor and a new graphical interface for easy setup and operation
  • Brushless motor offers 0.1% speed control accuracy with a 6000 to 1 turndown
  • Remote control capabilities include scaleable speed control input, start/stop/prime, motor direction, and tachometer output
  • Dispense by volume, time, or copy; cumulative volume function totalizes volume dispensed over multiple batches
  • Flow rates from 0.006 to 3400 mL/min with Masterflex L/S pump tubing
Kimble Screwhead Disposable Culture Tubes
Featured Product!
Kimble® Screwhead Disposable Culture Tubes
Ideal for general bacteriological and tissue culture applications
  • Manufactured of borosilicate glass
  • Tubes are offered with or without white markings
  • Phenolic screw caps feature a cemented-in rubber liner
  • Caps ordered separately
stir-pak mixer
Featured Product!
NEW Cole-Parmer Stir-Pak® General-Purpose Mixer
This general-purpose mixer is ideal for routine, light-duty mixing of water-like viscosities only delivering speeds from 0 to 4000 rpm.
The brushed DC motor has a compact speed controller integrated by a custom anodized aluminum extrusion. The potentiometer controls both on/off and variable speed settings for single direction mixing.
IKA Incubating Shakers-SAVE HUNDREDS ON NEW IKA SHAKING INCUBATORS
Featured Product!
IKA® Incubating Shakers
Reduced price—save hundreds on new IKA shaking incubators!
  • Advanced temperature control and safety features enable unattended operation in a temperature-controlled environment
  • Antimicrobial coating reduces bacteria on control surface
  • Easy-to-read LED display of speed, temperature, and time
  • All functions can be controlled and documented using the labworldsoft® software
Or visit our
New Product Page
Submit a product review today!
  • Visit ColeParmer.com
  • Special Offers!
  • Complete Online Catalog
  • Technical Articles and Buyer’s Guides
  • Request Free Catalogs
  • Give Us Your Opinion
    Information and supplies for
    pandemic flu
  • Ultrasonic Fractionation
    of Cells in Microplates
  • An Invaluable Tool for Water Analysis
  • What You Should Know about Temperature Control and Hot Plates
  • New Masterflex® L/S® Digital Drives
  • Kimble® Screwhead Disposable Culture Tubes
  • NEW Cole-Parmer Stir-Pak® General Purpose Mixer
  • Reduced price— on new IKA shaking incubators
    New 2009/10 Cole-Parmer General Catalog
    Lab Necessities Catalog
    2008 Water Quality Catalog
    2009 Cole-Parmer Mixers Catalog
    Spring 2009 Cole-Parmer Preferred Solutions Catalog
    Fittings Tubing Valves and Flowmeters Catalog
    Canadian customer click here
    Customers in the UK click here
    Customers in China click here
    Customers in India click here
    ©2009 Cole-Parmer. All rights reserved.
    625 East Bunker Ct. Vernon Hills, IL 60061-1844 USA
    This is an advertising message from Cole-Parmer.
    To ensure delivery, add Cole-Parmer to your address book.
    You can modify your profile or unsubscribe anytime.
    Your privacy is important to us—please review our online privacy policy.
    Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
    Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data