Cole-Parmer Technical Library
Flowmeter Selection Guide
When selecting a flow meter consideration must be given to:
  • Flow measurement type - momentum (velocity), volumetric or mass flow measurement
  • Media - type of media (liquid, gas or slurry) and any special condition such as particulates in the media and viscosity of the media
  • Media conditions - pressure and temperature of media and whether media conditions are likely to remain constant or vary
  • Flow range - required flow range of media (min and max readings required)
  • Accuracy - required accuracy of the readings
  • Environmental considerations - special installation considerations such as hygienic installation, installing in to an ATEX zone or requiring tamper proof readings
For special or unique flow control applications use the helpful flow meter application table below to narrow your selection.
Application Parameter Gases Liquids Flowmeter type
Low-flow measurement/control (some types as low as 1 sccm air and 0.1 mL/min water, max of range)
Mechanical orifice (controller)
Thermal dispersion
Coriolis
Differential pressure
Indicators/sensors/switches
Turbine(Pelton wheel)
Variable area
Gear
Propeller type
Magnetic
Ultrasonic
Vortex
High-flow measurement/control (some types as high as 2300 scfm gas and 2600 GPM liquid)
Thermal dispersion
Coriolis
Differential pressure
Indicators/switches
Variable area
Vortex
Propeller type
Magnetic
Turbine
Ultrasonic
High pressure (500 psi and up)
Mechanical orifice (controller)
Thermal dispersion
Coriolis
Indicators/switches
Variable area
Vortex
Differential pressure
Gear
Magnetic
Turbine
Ultrasonic
High temperature (200°F and up)
Mechanical orifice (controller)
Coriolis
Vortex (including steam)
Variable area
Ultrasonic
Turbine
Magnetic
Indicators/sensors
Propeller type
Gear
Differential pressure
High-accuracy measurement (error at or below 1%)
Differential pressure
Thermal dispersion
Coriolis
Vortex
Gear
Propeller type
Magnetic
Turbine (Pelton wheel)
Aggressive or pure fluids
Mechanical orifice (controller)
Thermal dispersion
Turbine
Variable area
Gear
Propeller type
Indicators/switches
Magnetic (aggressive only)
Ultrasonic
Vortex
Fluids with heavy particulates or slurries
Coriolis
Magnetic
Ultrasonic
High-viscosity fluids
Coriolis
Gear
Magnetic
No wired power available
Mechanical orifice (controller)
Differential pressure
Indicators
Turbine
Variable area
Gear
Propeller type
Ultrasonic
Intrinsically safe
Mechanical orifice (controller)
Variable area
Differential pressure
Gear
Propeller type
Indicators
Turbine
Large pipe sizes (3" or larger)
Coriolis
Vortex
Ultrasonic
Turbine (insertion)
Magnetic
Propeller type
Sanitary (meets 3A standards)
Coriolis (special order)
Magnetic
Turbine
Related Links: Printable page E-mail a colleague
Complete Selection of Flowmeters
Flowmeter Applications
How Volumetric Flowmeters Work
Installing Your Paddle-Wheel Flow Sensor
Selecting the Right Flowmeter—Part 1
Selecting the Right Flowmeter—Part 2
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data