Youre serious about your electrical
test instruments. You buy top
brands, and you expect them to be
accurate. You know some people
send their digital instruments to a
metrology lab for calibration, and
you wonder why. After all, these
are all electronic theres no
meter movement to go out of balance.
What do those calibration
folks do, anyhow just change
the battery?
These are valid concerns,
especially since you cant use your
instrument while its out for calibration.
But, lets consider some
other valid concerns. For example,
what if an event rendered your
instrument less accurate, or
maybe even unsafe? What if you
are working with tight tolerances
and accurate measurement is key
to proper operation of expensive
processes or safety systems? What
if you are trending data for maintenance
purposes, and two meters
used for the same measurement
significantly disagree?
Many people do a field comparison check of two
meters, and call them calibrated if they give the
same reading. This isnt calibration. Its simply a
field check. It can show you if theres a problem,
but it cant show you which meter is right. If both
meters are out of calibration by the same amount
and in the same direction, it wont show you anything.
Nor will it show you any trending you
wont know your instrument is headed for an out
of cal condition.
For an effective calibration, the calibration standard
must be more accurate than the instrument
under test. Most of us have a microwave oven or
other appliance that displays the time in hours and
minutes. Most of us live in places where we
change the clocks at least twice a year, plus again
after a power outage. When you set the time on
that appliance, what do you use as your reference
timepiece? Do you use a clock that displays seconds?
You probably set the time on the digitschallenged
appliance when the reference clock is
at the top of a minute (e.g., zero seconds). A
metrology lab follows the same philosophy. They
see how closely your whole minutes track the
correct number of seconds. And they do this at
multiple points on the measurement scales.
Calibration typically requires a standard that
has at least 10 times the accuracy of the instrument
under test. Otherwise, you are calibrating
within overlapping tolerances and the
tolerances of your standard render an in cal
instrument out of cal or vice-versa. Lets look at
how that works.
Two instruments, A and B, measure
100 V within 1 %. At 480 V, both are
within tolerance. At 100 V input, A reads
99.1 V and B reads 100.9 V. But if you
use B as your standard, A will appear to
be out of tolerance. However, if B is
accurate to 0.1 %, then the most B will
read at 100 V is 100.1 V. Now if you
compare A to B, A is in tolerance. You
can also see that A is at the low end of
the tolerance range. Modifying A to bring
that reading up will presumably keep A
from giving a false reading as it experiences
normal drift between calibrations.
Calibration, in its purest
sense, is the comparison of an
instrument to a known standard.
Proper calibration involves use of
a NIST-traceable standard one
that has paperwork showing it
compares correctly to a chain of
standards going back to a master
standard maintained by the
National Institute of Standards
and Technology.
In practice, calibration
includes correction. Usually
when you send an instrument for
calibration, you authorize repair
to bring the instrument back into
calibration if it was out of cal.
Youll get a report showing how
far out of calibration the instrument
was before, and how far
out it is after. In the minutes and
seconds scenario, youd find the
calibration error required a correction
to keep the device dead
on, but the error was well
within the tolerances required
for the measurements you made
since the last calibration.
If the report shows gross calibration
errors, you may need to
go back to the work you did with
that instrument and take new
measurements until no errors are
evident. You would start with the
latest measurements and work
your way toward the earliest
ones. In nuclear safety-related
work, you would have to redo all
the measurements made since
the previous calibration.
What knocks a digital instrument
out of cal? First, the major
components of test instruments
(e.g., voltage references, input
dividers, current shunts) can
simply shift over time. This shifting
is minor and usually harmless
if you keep a good calibration
schedule, and this shifting is typically
what calibration finds and
corrects.
But, suppose you drop a current
clamp hard. How do you
know that clamp will accurately
measure, now? You dont. It may
well have gross calibration
errors. Similarly, exposing a DMM
to an overload can throw it off.
Some people think this has little
effect, because the inputs are
fused or breaker-protected. But,
those protection devices may not
trip on a transient. Also, a large
enough voltage input can jump
across the input protection
device entirely. This is far less
likely with higher quality DMMs,
which is one reason they are
more cost-effective than the less
expensive imports.
The question isnt whether to
calibrate we can see thats a
given. The question is when to
calibrate. There is no one size
fits all answer. Consider these
calibration frequencies:
- Manufacturer-recommended
calibration interval. Manufacturers
specifications will
indicate how often to calibrate
their tools, but critical measurements
may require different
intervals.
- Before a major critical measuring
project. Suppose you are
taking a plant down for testing
that requires highly accurate
measurements. Decide which
instruments you will use for
that testing. Send them out for
calibration, then lock them
down in storage so they are
unused before that test.
- After a major critical measuring
project. If you reserved
calibrated test instruments for
a particular testing operation,
send that same equipment for
calibration after the testing.
When the calibration results
come back, you will know
whether you can consider that
testing complete and reliable.
- After an event. If your instrument
took a hit something
knocked out the internal overload
or the unit absorbed a
particularly sharp impact
send it out for calibration and
have the safety integrity
checked, as well.
- Per requirements. Some
measurement jobs require
calibrated, certified test equipment
regardless of the
project size. Note that this
requirement may not be
explicitly stated but simply
expected review the specs
before the test.
- Monthly, quarterly, or semiannually.
If you do mostly
critical measurements and do
them often, a shorter time span
between calibrations means
less chance of questionable test
results.
- Annually. If you do a mix of
critical and non-critical measurements,
annual calibration
tends to strike the right balance
between prudence and cost.
- Biannually. If you seldom do
critical measurements and dont
expose your meter to an event,
calibration at long frequencies
can be cost-effective.
- Never. If your work requires
just gross voltage checks (e.g.,
Yep, thats 480V), calibration
seems like overkill. But what if
your instrument is exposed to
an event? Calibration allows
you to use the instrument with
confidence.