  Cart  Subtotal = 0.00
1-888-358-4717 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Cole-Parmer Technical Library
Mission Impossible:
Measuring the pH of Pure Water!
You wouldn't think it would be so darn difficult!
By Frank Paparone, Technical Support Leader, OAKTON Instruments
What could possibly be so difficult about reading the pH of pure water? It should be neutral—pH 7.0—without any ion interference, right? In fact, it can be quite difficult and often frustrating to obtain reproducible pH values in samples with low-ionic strength.
Water that has very few ionic species is said to be low in alkalinity, ionic strength, or to have low conductivity/high resistivity, such as with distilled or deionized (DI) water. And due to the varying junction potentials that develop across the reference junction, when attempting to measure DI water it is common to attain different pH values even with new, sealed electrodes that calibrate perfectly in pH buffers.
Choosing a pH Electrode
More expensive double-junction and calomel electrodes have a number of advantages over traditional electrodes. However, they are generally no better than their counterparts for pure water applications. The best choice is a refillable, liquid-filled electrode, ideally made of low resistance glass. A flowing reference junction has a higher flow rate to minimize junction potentials. Sealed electrodes, usually gel-filled, are best known for their "long-life", due in part to the extremely slow leak rate of the reference solution. However, a fast leak rate is desirable with pure water so that a pH potential can be established more quickly. This is also the reason why sealed electrodes should not be stored in DI water—the sample will accelerate the leaching of reference solution, which cannot be replaced.
Electrostatic Interference
More Details or Order Online:
OAKTON® pH/Con/TDS Benchtop Meter
Refillable Glass-Body Combination Electrode
Since ultra pure water is a bad conductor, it can also be a source of static potentials. These static potentials may present problems while measuring pH. To compensate for this interference, pH electrodes are available with a special shielded or grounded compartment. Such electrodes are expensive and specifically designed for measurement of ultra pure water.
Other Measurement Tips
  • It is also beneficial to measure pH in the smallest sample volume possible. Direct pH measurement in large volume samples, such as drums or tanks and other samples with flowing or moving water, produces results that tend to fluctuate and will require excessive stabilization time.
  • Addition of a tiny amount of KCl will increase ionic strength to the sample and improve response time. However, only high purity KCl should be used as trace contaminants, as low-grade KCl can artificially alter the pH.
  • Maintain good laboratory practices (GLP), including using clean glassware, avoiding cross-contamination after calibration by rinsing thoroughly with DI water, and using only certified calibration standards.
  • Temperature compensation should be used during measurement and calibration. Calibrate daily at multiple pH calibration points.
  • Minimize exposure of your sample with air. CO2 gas absorption can actually decrease pH.
Related Links: Printable page E-mail a colleague
Complete Selection of pH Electrodes
Complete Selection of pH/Ion Meters
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data