| Cole-Parmer Technical Library | |
|
|
|
The Bostwick Consistometer consists of a level, stainless-steel trough with two compartments. The first compartment, which holds the sample to be tested, is separated from the second compartment by a spring-loaded gate. The second compartment is 24 cm long and has graduated, parallel lines at 0.5 cm intervals. The measurement consists of releasing the gate and letting the fluid flow freely under gravity down the slope. The distance the fluid flows from the gate after 30 seconds is measured in centimeters as the Bostwick Consistometer reading. Bostwick LimitationsThe Bostwick Consistometer presents two problems. First, the instrument is limited in the range of products it can test. For example, it is not suitable for high-solids tomato paste because the paste does not flow far enough in 30 seconds to give measurable differences between samples. Secondlyand more importantlythe results from this instrument cannot be converted into fundamental rheological parameters because surface tension, wetting power and factors other than viscosity may also be involved. There is also no way to control sample temperature. Rotational Technology
The rotational viscometer measures viscosity by determining the viscous resistance of the fluid. This measurement is obtained by immersing a spindle into the test fluid. The viscometer measures the additional torque required for the spindle to overcome viscous resistance and regain constant speed. This value is then converted to centipoise and displayed on the instrument's LCD readout. Testing Tomato Sauce
Rotational viscometers are a necessity for any contemporary high-tech food laboratory.
Complete Selection of Brookfield Rotational Viscometers |
Product Selection Guides | Product Maintenance | FAQs | Technical Library Map
Articles and White Papers | Case Studies | Conversion Factors and Tech Data