General Pump FAQs
- How do pumps work?
- Are centrifugal pumps variable speed?
- What exactly is a positive displacement pump?
- Which pumps can I run dry?
- What is the maximum viscosity rating for pumps?
- What pumps do you carry that will handle particulates?
- I need gentle pumping action, what do you recommend?
- When do you need to perform maintenance on pumps?
Pumps move fluid in a variety of ways:
-
- Use centrifugal force to push the fluid through the outlet.
-
- Bellows, diaphragm, peristaltic, piston, and syringe pumps are all metering pumps that pull the fluid through the inlet valve into a chamber, close the inlet valve, and then push the fluid through the outlet valve.
- - Bellows, double-diaphragm, flexible impeller, gear, oscillating, piston, progressing cavity, rotary lobe, rotary vane, and peristaltic pumps have a fixed cavity that the fluid is pushed through by rollers, gears, or impeller. As the fluid is pushed through, it leaves a void or vacuum which pulls in more fluid.
Go to Top
Most centrifugal pumps do not have variable speed motors. However, you can control
flow rate on the discharge using a valve.
Go to Top
A positive displacement pump emits a given volume of fluid for each revolution of
the motor. Bellows, double-diaphragm, flexible impeller, gear, oscillating, piston,
progressing cavity, rotary lobe, rotary vane, and peristaltic pumps are all positive displacement pumps.
Go to Top
Peristaltic, piston pumps with ceramic heads, bellows pumps, and diaphragm pumps
can be run dry for any length of time. Centrifugal, rotary vane, and gear pumps should not
be run dry; exceptions are if the gear or impeller is made of a self-lubricating material
such as RYTON in which case the pump can be run for a few minutes while priming.
Go to Top
This depends on the type of pump and the specific pump. Diaphragm pumps
(especially double diaphragm pumps) and gear pumps are usually the best for viscous fluids.
Go to Top
Diaphragm pumps, bellows pumps and peristaltic pumps will work well. When choosing
materials, consider chemical compatibility and resistance to wear. Use a pump with larger
fittings so they don't clog as easily.
Go to Top
A peristaltic pump, used at low speeds. You can also use a diaphragm pump, again
at low speed. Centrifugal and gear pumps, which work at high speeds and have high shear rates,
should be avoided.
Go to Top
This depends on the pump and the application. In general, diaphragms on metering
pumps last about 6 to 12 months; gears on gear pumps last about 3 to 6 months; and motors
usually last for years. DC motors require periodic brush replacement. It is important to
monitor brush wear; normally brushes should be replaced every 6 months.
Go to Top
Complete Selection of Pumps - Centrifugal Pumps Complete Selection of Pumps - Drum and Hand Pumps Complete Selection of Pumps - Gear Pumps Complete Selection of Pumps - Ismatec Complete Selection of Pumps - Manostat Peristaltic Pumps Complete Selection of Pumps - Masterflex Peristaltic Pumps Complete Selection of Pumps - Metering Pumps Complete Selection of Pumps - Postive Displacement Pumps
General Pump FAQs
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Terminology
Liquid Pumps Technical Equations and Information Tables
Pump Types and Definitions