  Cart  Subtotal = 0.00
1-888-358-4717 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Cole-Parmer Technical Library
Weighing Unit Conversion Table
To convert from grams to: Abbreviation multiply by
carats ct 5.0
dram 0.564383
grains gn 15.432358
momme mom 0.2666666
ounces oz 0.0352739
pennyweights dwt 0.6430149
pounds lb 0.0022046
scruple 0.7716179
tael (Hong Kong) tlh 0.0264554
tola (India) tol 0.0857353
tael (Singapore) tls 0.0264599
tael (Taiwan) tlt 0.0266666
troy ounces oz t 0.0321507
troy pounds lb t 0.0026792
Related Links: Printable page E-mail a colleague
Complete Selection of Balances
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data