Cole-Parmer Technical Library
Technical Equations
Centrifugal Force    Concentration     Flow Heating and Cooling    Humidity    
Viscosity     Ohm's Law
Centrifugal Force
Relative centrifugal force
rcf = (11.18 x 10-6) RN
R = rotating radius in cm
N = rotation speed in rpm
Concentration
Molar (M) =
Moles of solute
Liters of solution
Weight % =
     Grams of solute x 100%     
Grams of solute + grams of solvent
Volume % =
Liters of solute x 100%
Liters of solution
ppm =
mg of solute
kg of solution
=
         mg        
Liters of water
Flow
CV factor
Q =
     CV     
(G/ΔP)1/2
Q = flow rate (GPM)
CV = flow coefficient
G = specific gravity
ΔP = pressure drop (psi)
Heating and Cooling
Cooling capacity of chillers
Btu/hr =
ΔT (°F) x Specific heat of fluid
flow rate (GPM)
Specific heat of water = 500 Btu/hr/GPM/°F
Specific heat of oil = 350 Btu/hr/GPM/°F
Heating water in tanks
kW =
Volume (gallons) x ΔT (°F)
325 x Heat-up time (hours)
Heating oil in tanks
kW =
Volume (gallons) x ΔT (°F)
800 x Heat-up time (hours)
Heating flowing water
kW = Flow (GPM) x ΔT (°F) x 0.16
Humidity
Absolute humidity (as g/m3)
D =
        804        
1 + 0.00633t
x
   e   
PO
D =
   H   
100
x
        804        
1 + 0.00633t
x
   e   
PO
Dew point temperature (as °C)
T =
237.3/7.5 - 1
log e - 0.786
e =
   H   
100
x eS
eS = 6.1078 x 107.5t/(237.3 + t)
Relative humidity (as % RH)
RH =
  e  
eS
x 100 =
  D  
DS
x 100
H = relative humidity (% RH)
PO = standard air pressure (mm Hg)
D = absolute humidity (g/m3)
e = water steam pressure (mm Hg)
DS = absolute humidity in saturation (g/m3)
eS = saturated water steam pressure (mm Hg)
t = temperature (°C)
T = dew point temperature
Viscosity
Absolute viscosity = kinematic viscosity x density
density = units of specific gravity or g/mL
(eg. poise = stokes x g/mL)
Ohm's Law
Volts
Volts = √Watts x Ohms =
  Watts  
Amperes
= Amperes x Ohms
Amperes
Amperes =
Volts
Ohms
=
Watts
Volts
= √
Watts
Ohms
Ohms
Ohms =
    Volts    
Amperes
=
Volts2
Watts
=
    Watts    
Amperes2
Watts
Watts = Volts x Amperes = Amperes2 x Ohms =
Volts2
Ohms
Related Links: Printable page E-mail a colleague
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data