  Cart  Subtotal = 0.00
1-888-358-4717 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Contact Us
Find a Global Dealer
Career Opportunities
About Us
Mission Statement
Our History
Milestones
Benefits
Directions to Cole-Parmer
Business Profile
Quality Policy
Executive Team
Environmental, Safety, and Health Policy
Trademarks
Press Releases
Event Calendar
FeedBack
Help
Cole-Parmer Technical Library
Chemical Compatibility Chart
For Disposable Gloves
A Warning: The information in this chart was supplied to Cole-Parmer by other reputable sources and is to be used ONLY as a guide in selecting gloves for chemical compatibility testing. Variability in material thickness, chemical concentration, temperature, and length of exposure to chemicals will affect specific performance.
Always test the products with the specific chemicals and under the specific conditions of your application. Cole-Parmer does not warrant (neither expressed nor implied) the accuracy or completeness of this chart or that any material is suitable for any purpose.
Compatibility Ratings
A — Very Good or Excellent
B — Good
C — Fair
D — Not recommended
— No data available
Brand of glove Microflex® Ansell Best™
N-DEX®
Diamond Grip™
Plus
NeoPro™ Nitron One® Freeform™ Touch-N-Tuff™
Glove material Latex Chloroprene Nitrile Nitrile Nitrile Nitrile
Chemical Compatibility rating
Acetic Acid A A A A A D
Acetone A B D D D D
Acetonitrile A C D
Ammonium Hydroxide A A A A B A
Carbon Tetrachloride D C B B D
Chloroform D B D D D
Ethanol A A A A D
Ethyl Ether B A B B B D
Formaldehyde A A A A A A
Hexane D C B B A A
Hydrochloric Acid B A B B A A
Isopropanol A A
Kerosene C A A A A
Methanol A A A A A
Nitric Acid; 10% D B C C A A
Phosphoric Acid B A A A A
Potassium Hydroxide A A A A A
Sodium Hydroxide A A A A A A
Sulfuric Acid; 47% Battery Acid A B B B A B
Related Links: Printable page E-mail a colleague
Complete Selection of Gloves
Chemical Compatibility Chart for Reusable Gloves
Safety Gloves Selection Guide
Safety Gloves Sizing Guide
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data