Environmental Express® BOD Testing
Environmental Express offers a variety of testing products for Biochemical Oxygen Demand (BOD) analysis. From our pre-measured SimpleWater Nutrient Buffer Solution to our one-of-a-kind disposable BOD bottles, our products prove to be efficient and cost-effective.
There is no competition—we manufacture the world’s only eco-conscious single-use BOD bottle available for EPA and Standard Methods. Our patent pending colored BOD stoppers allow you to segment and visually identify your samples with ease.
BOD product group
Disposable & Glass Biochemical Oxygen Demand (B.O.D.) Bottles
View All BOD Bottles >
Environmental Express BOD bottles provide huge labor savings and improved quality in any environmental laboratory. Lightweight, unbreakable and recyclable, disposable bottles offer safety as well as convenience.
The US EPA recommends the use of Environmental Express BOD bottles be allowed for EPA approved Methods.
Click here for US EPA Letter of Approval Click here for an independent comparative study on glass bottles versus disposable.
Bottles are formed from PET resin, the most recycled plastic available. Simply use once and toss into the recycling bin. Consider how many gallons of water per year you use washing, triple rinsing and DI rinsing your glass BOD bottles. With eco-conscious single-use bottles you conserve water and eliminate waste water treatment.
The use of clean, new bottles eliminates the problem of contamination from sample carry over or detergent residue. Our disposable bottles have an amorphous carbon coating which keeps oxygen from diffusing into or out of your sample. Bottles provide uniform blanks, high standard recoveries and quality test data.
Bottles come packaged and are individually barcoded and numbered.
BOD Method Guide
Color Coded BOD Stopper | Easily Identify Your BOD Bottle Contents
View All BOD Stoppers > and BOD Overcaps >
Environmental Express plastic BOD stoppers work with all brands of glass and plastic, 60mL and 300mL BOD bottles. Easily identify what's in the BOD bottle based on the color of the stopper. The colored cap on the BOD stopper can be used to differentiate your samples as shown below or you can create your own plan:
Blue (D1025-BLUE) - samples and duplicates
Black (D1025-BLACK) - blanks
Green (D1025-GREEN) - quality control (GGA) samples
Red (D1025-RED) - seed control factor
Speak to your sales representative about custom color options (minimum purchase quantities may apply).
*BOD Stopper Patent Pending
BOD Incubators
View All Refrigerated Incubators >
BOD incubator
BOD Racks and Carriers
View BOD Bottle Storage Racks and Carriers >
BOD rack
BOD Reagents
View BOD Dilution Water Solution Bottle Sets >
View SimpleWater BOD Nutrient Buffer Solution >
View Polyseed Capsules for BOD and CBOD >
Dilution water
Seed
BOD Standards
View Single-Use Glucose-Glutamic Acid (GGA) Vials >
BOD standard
DO Meters
View All Dissolved Oxygen Meter Products >
DO meters
Water Purification Systems
Thermo Scientific Barnstead 50129885 Smart2Pure, 6 l/h, UV >
Thermo Scientific Barnstead 50132373 MicroPure water system, UV >
Water purification system